Skip to Content
Merck
All Photos(1)

Key Documents

488712

Sigma-Aldrich

Trimethyl(trifluoromethyl)silane

99%

Synonym(s):

(Trifluoromethyl)trimethylsilane, Ruppert’s Reagent, TFMTMS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCF3
CAS Number:
Molecular Weight:
142.19
Beilstein:
4241868
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

reaction suitability

reaction type: C-C Bond Formation

bp

54-55 °C (lit.)

density

0.962 g/mL at 20 °C (lit.)

functional group

fluoro

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C(F)(F)F

InChI

1S/C4H9F3Si/c1-8(2,3)4(5,6)7/h1-3H3

InChI key

MWKJTNBSKNUMFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethyl(trifluoromethyl)silane can be used as a trifluoromethylating agent in the following processes:
  • Conversion of N-(tert-butylsulfinyl)-imines to trifluoromethylated amines
  • Conversion of trans-enones to trans-α-trifluoromethyl silyl ethers
  • Trifluoromethylation of azomethine imines
  • Conversion of H-phosphonates to CF3-phosphonates
  • Nucleophilic addition of the trifluoromethyl group to aldehydes and ketones.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-catalyzed aerobic oxidative trifluoromethylation of H-phosphonates using trimethyl (trifluoromethyl) silane
Chu L, et al.
Synthesis, 44(10), 1521-1525 (2012)
Stereoselective Nucleophilic Trifluoromethylation of N?(tert?Butylsulfinyl) imines by Using Trimethyl (trifluoromethyl) silane.
Prakash G K, et al.
Angewandte Chemie (International Edition in English), 113(3), 609-610 (2001)
Peter T Kaplan et al.
Beilstein journal of organic chemistry, 13, 2297-2303 (2017-11-29)
A number of copper reagents were compared for their effectiveness in trifluoromethylating 4-iodobiphenyl, 4-iodotoluene, and 2-iodotoluene. Yields over time were plotted in order to refine our understanding of each reagent performance, identify any bottlenecks, and provide more insight into the
Catalytic enantioselective trifluoromethylation of azomethine imines with trimethyl (trifluoromethyl) silane
Kawai H, et al.
Angewandte Chemie (International Edition in English), 121(34), 6442-6445 (2009)
CsF-catalyzed nucleophilic trifluoromethylation of trans-enones with trimethyl (trifluoromethyl) silane: A facile synthesis of trans-?-trifluoromethyl allylic alcohols.
Singh R P, et al.
Organic Letters, 1(7), 1047-1049 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service