Skip to Content
Merck
All Photos(2)

Key Documents

217220

Sigma-Aldrich

Ytterbium(III) nitrate pentahydrate

99.999%

Synonym(s):

Ytterbium(+3) trinitrate pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Yb(NO3)3 · 5H2O
CAS Number:
Molecular Weight:
449.13
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999%
form:
crystals, granules or chunks

Quality Level

Assay

99.999%

form

crystals, granules or chunks

reaction suitability

reagent type: catalyst
core: ytterbium

impurities

≤15.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.[Yb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.5H2O.Yb/c3*2-1(3)4;;;;;;/h;;;5*1H2;/q3*-1;;;;;;+3

InChI key

XIOPWXFTXDPBEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ytterbium(III) nitrate pentahydrate is a white hygroscopic salt. It is soluble in water and ethanol. It is widely used for the synthesis of ceramics.

Application

Ytterbium(III) nitrate pentahydrate can be used:
  • As a precursor to prepare Tb3+–Yb3+ co-doped CaF2 phosphors prepared by combustion synthesis.
  • As a dopant to synthesize Cs2AgInCl6 double perovskite with optical functionality in the NIR(near infrared) window.
It forms superhydrophobic ceramic coating by using Solution Precursor Plasma Spray.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Near-Infrared Emission of Yb-Doped Cs2AgInCl6 Double Perovskite Microcrystals and Nanocrystals
Yogesh Mahor, et al.
The Journal of Physical Chemistry C, 123, 15787-15793 (2019)
Photon up-conversion production in Tb3+?Yb3+ co-doped CaF2 phosphors prepared by combustion synthesis
Nikifor Rakov, et al.
Materials Research Bulletin, 74, 103-108 (2016)
Superhydrophobic ceramic coatings by solution precursor plasma spray
Yuxuan Cai, et al.
Scientific Reports, 6, 24670-24670 (2016)

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service