Skip to Content
Merck
All Photos(2)

Key Documents

215260

Sigma-Aldrich

Rubidium chloride

99.8% trace metals basis

Synonym(s):

Rubidium monochloride, Rubidium(I) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbCl
CAS Number:
Molecular Weight:
120.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

Assay

99.8% trace metals basis

form

powder and chunks

impurities

≤2500.0 ppm Trace Rare Earth Analysis

mp

715 °C (lit.)

density

2.8 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[Rb+]

InChI

1S/ClH.Rb/h1H;/q;+1/p-1

InChI key

FGDZQCVHDSGLHJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

RbCl, an alkali halide may be used to dope L-alanine hydrogen chloride monohydrate single crystals to alter its dielectric properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Growth, structural, spectral, mechanical and dielectric characterization of RbCl-doped l-alanine hydrogen chloride monohydrate single crystals
Rose AL, et al.
Physica B: Condensed Matter, 406.3, 412-417 (2011)
Balasundaresan Dhakshnamoorthy et al.
Journal of molecular biology, 396(2), 293-300 (2009-11-26)
The OmpF porin from the Escherichia coli outer membrane folds into a trimer of beta-barrels, each forming a wide aqueous pore allowing the passage of ions and small solutes. A long loop (L3) carrying multiple acidic residues folds into the
Eiichi Takayama et al.
Journal of molecular and cellular cardiology, 37(2), 483-496 (2004-07-28)
We previously demonstrated that pinacidil does not affect Na(+)(i) accumulation, cellular energy depletion, or acidosis during myocardial ischemia, but dramatically improves the cationic/energetic status during reperfusion. We investigated the role of this latter effect in K(ATP) channel-induced cardioprotection. Employing (23)Na
Takuro Hayashi et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 65(Pt 10), 1007-1010 (2009-10-24)
Small heat-shock proteins (sHsps) bind and stabilize proteins denatured by heat or other stresses in order to prevent unfavourable protein aggregation. StHsp14.0 is an sHsp found in the acidothermophilic archaeon Sulfolobus tokodaii. A variant of StHsp14.0 was crystallized by the
Amer Alam et al.
Nature structural & molecular biology, 16(1), 30-34 (2008-12-23)
We report the crystal structure of the nonselective cation channel NaK from Bacillus cereus at a resolution of 1.6 A. The structure reveals the intracellular gate in an open state, as opposed to the closed form reported previously, making NaK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service