Skip to Content
Merck
All Photos(3)

Key Documents

204250

Sigma-Aldrich

Rubidium chloride

99.95% trace metals basis

Synonym(s):

Rubidium monochloride, Rubidium(1+)chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbCl
CAS Number:
Molecular Weight:
120.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.95% trace metals basis
form:
powder and chunks

Quality Level

Assay

99.95% trace metals basis

form

powder and chunks

impurities

≤550.0 ppm Trace Metal Analysis

mp

715 °C (lit.)

density

2.8 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[Rb+]

InChI

1S/ClH.Rb/h1H;/q;+1/p-1

InChI key

FGDZQCVHDSGLHJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • A Study on the Removal of Impurity Elements Silicon and Zinc from Rubidium Chloride by Vacuum Distillation: This research focuses on the purification of rubidium chloride from impurities like silicon and zinc, which is crucial for maintaining the quality and effectiveness of this compound in various applications (Cui et al., 2024).
  • Effect of a Rubidium Chloride Carrier Confinement Layer on the Characteristics of CsPbBr3 Perovskite Light-Emitting Diodes: This article explores the impact of rubidium chloride as a confinement layer in light-emitting diodes, enhancing the performance of these devices, relevant for chemists working on advanced materials and electronic devices (Li et al., 2022).
  • Rubidium chloride modulated the fecal microbiota community in mice: This research assesses the effects of rubidium chloride on the fecal microbiota in mice, providing insights that could be useful for studies related to gut health and its impact on overall well-being (Chen et al., 2021).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yucheng Jiang et al.
Biophysical chemistry, 118(1), 25-30 (2005-07-26)
Electromotive force (emf) of the chemical cell without liquid-junction K-ISE | RbCl/CsCl ( m(E)) | ISE-Cl and K-ISE | RbCl/CsCl (m(E)), amide (m(N)) | ISE-Cl, have been measured at 298.15 K, where m(E)=(0.005 to 0.5) mol kg(-1) and m(N)= (0.05
Ferenc Horkay et al.
The Journal of chemical physics, 125(23), 234904-234904 (2006-12-28)
The distribution of counterions in solutions of high molecular mass hyaluronic acid, in near-physiological conditions where mono- and divalent ions are simultaneously present, is studied by small angle neutron scattering and anomalous small angle x-ray scattering. The solutions contain either
M L Główka et al.
Acta crystallographica. Section D, Biological crystallography, 61(Pt 4), 433-441 (2005-04-05)
Gramicidin D (gD) is a naturally occurring ionophoric antibiotic that forms membrane channels specific for monovalent cations. The crystal structure of the RbCl complex of gD has been determined at 1.14 A resolution from low-temperature (100 K) synchrotron-radiation data with
Grazyna Bystrzejewska-Piotrowska et al.
Journal of environmental radioactivity, 99(7), 1185-1191 (2008-03-18)
Ex vitro cultures of Pleurotus eryngii were carried out under controlled conditions using sterile medium composed of barley seeds. The influence of alkali and alkaline earth element salts (CsCl, KCl, NaCl, RbCl, and CaCl(2)) and tetraethylammonium chloride on incorporation of
Amer Alam et al.
Nature structural & molecular biology, 16(1), 30-34 (2008-12-23)
We report the crystal structure of the nonselective cation channel NaK from Bacillus cereus at a resolution of 1.6 A. The structure reveals the intracellular gate in an open state, as opposed to the closed form reported previously, making NaK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service