Skip to Content
Merck
All Photos(1)

Key Documents

213950

Sigma-Aldrich

Vinyltrimethylsilane

97%

Synonym(s):

Trimethyl(vinyl)silane, (Trimethylsilyl)ethylene, Ethenyltrimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCHCH2
CAS Number:
Molecular Weight:
100.23
Beilstein:
956572
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

4.44 psi ( 20 °C)

Assay

97%

form

liquid

impurities

<3% tetrahydrofuran

refractive index

n20/D 1.391 (lit.)

bp

55 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)C=C

InChI

1S/C5H12Si/c1-5-6(2,3)4/h5H,1H2,2-4H3

InChI key

GCSJLQSCSDMKTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used for the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-11.2 °F

Flash Point(C)

-24 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seco-steroids. I. 15,16-Secoprogesterone.
N S Crossley
Journal of the Chemical Society. Perkin transactions 1, 13, 2491-2495 (1971-01-01)
Synthesis, 1315-1315 (2007)
J. Chem. Soc. Perkin Trans. II, 1129-1129 (1993)
J. Mol. Catal., 90, 125-125 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service