All Photos(1)
About This Item
Linear Formula:
FC6H4CH2COCH3
CAS Number:
Molecular Weight:
152.17
Beilstein:
1936697
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95% (GC)
form
liquid
refractive index
n20/D 1.496 (lit.)
bp
106-107 °C/18 mmHg (lit.)
density
1.139 g/mL at 25 °C (lit.)
functional group
fluoro
ketone
SMILES string
CC(=O)Cc1ccc(F)cc1
InChI
1S/C9H9FO/c1-7(11)6-8-2-4-9(10)5-3-8/h2-5H,6H2,1H3
InChI key
ZUEKIIWSVFBTCM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
195.8 °F - closed cup
Flash Point(C)
91 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service