Skip to Content
Merck
All Photos(2)

Documents

B90203

Sigma-Aldrich

Butylbenzene

≥99%

Synonym(s):

1-Phenylbutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)3CH3
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903395
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

vapor pressure

1.03 mmHg ( 23 °C)

Assay

≥99%

form

liquid

autoignition temp.

774 °F

expl. lim.

5.8 %

refractive index

n20/D 1.489 (lit.)

bp

183 °C (lit.)

mp

−88 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

CCCCc1ccccc1

InChI

1S/C10H14/c1-2-3-7-10-8-5-4-6-9-10/h4-6,8-9H,2-3,7H2,1H3

InChI key

OCKPCBLVNKHBMX-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Butylbenzene undergoes oxidation to afford butyrophenone. It is used to prepare N-arylazoles via oxidant-free and selective C(sp2)-H amination reaction. It can be used in the synthesis of alkylated pentacene and ladder-type oligo(p-phenylene)s to improve solubility in common organic solvents.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Study on liquid crystallinity in 2, 9?dialkylpentacenes.
Okamoto K , et al.
Liq. Cryst., 34(9), 1001-1007 (2007)
Photo-induced oxidant-free oxidative C?H/N?H cross-coupling between arenes and azoles.
Niu L, et al.
Nature Communications, 8, 14226-14226 (2017)
A New Method for the Benzylic Oxidation of Alkylarenes Catalyzed by Hypervalent Iodine (III).
Xu Y, et al.
Synthesis, 45(03), 370-374 (2013)
Ladder-type oligo (p-phenylene) s with D???A architectures: design, synthesis, optical gain properties, and stabilized amplified spontaneous emission.
Fang M, et al.
Journal of Material Chemistry C, 5(23), 5797-5809 (2017)
Dirhodium (II) Complexes of 2?(Sulfonylimino) pyrrolidine: Synthesis and Application in Catalytic Benzylic Oxidation.
Wusiman A, et al.
European Journal of Organic Chemistry, 2012(16), 3088-3092 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service