Skip to Content
Merck
All Photos(1)

Key Documents

202142

Sigma-Aldrich

Cesium bromide

99.9% trace metals basis

Synonym(s):

Cesium monobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsBr
CAS Number:
Molecular Weight:
212.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
crystalline

Quality Level

Assay

99.9% trace metals basis

form

crystalline

impurities

≤1500.0 ppm Trace Metal Analysis

mp

636 °C (lit.)

density

4.44 g/mL at 25 °C (lit.)

SMILES string

[Br-].[Cs+]

InChI

1S/BrH.Cs/h1H;/q;+1/p-1

InChI key

LYQFWZFBNBDLEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cesium bromide can be used:
  • As a precursor to synthesize cesium lead bromide nanoparticles by the ultrasonic bath method.
  • To fabricate inorganic perovskite material such as (CsPbBr3), for solar cells.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tatiana G Liashenko et al.
Nanomaterials (Basel, Switzerland), 10(10) (2020-10-03)
Inexpensive perovskite light-emitting devices fabricated by a simple wet chemical approach have recently demonstrated very prospective characteristics such as narrowband emission, low turn-on bias, high brightness, and high external quantum efficiency of electroluminescence, and have presented a good alternative to
Jingwei Zhu et al.
ChemSusChem, 13(7), 1834-1843 (2020-01-24)
The preparation of high-quality perovskite films with low grain boundaries and defect states is a prerequisite for achieving high-efficiency perovskite solar cells (PSCs) with good environmental stability. An effective additive engineering strategy has been developed for simultaneous defect passivation and
Olga Nazarenko et al.
Inorganic chemistry, 56(19), 11552-11564 (2017-09-13)
Interest in hybrid organic-inorganic lead halide compounds with perovskite-like two-dimensional crystal structures is growing due to the unique electronic and optoelectronic properties of these compounds. Herein, we demonstrate the synthesis, thermal and optical properties, and calculations of the electronic band
Cong Chen et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(11), 1802046-1802046 (2019-06-11)
Photovoltaic devices employing lead halide perovskites as the photoactive layer have attracted enormous attention due to their commercialization potential. Yet, there exists challenges on the way to the practical use of perovskite solar cells (PSCs), such as light stability and
Peng Pan et al.
Analytical chemistry, 75(20), 5468-5474 (2004-01-09)
Solutions consisting of protein and small molecule mixtures have been subjected to electrospray ionization to study the influence of small molecule/cation components at high concentrations on the electrospray responses of proteins. Emphasis was placed on solutions consisting of equal parts

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service