Skip to Content
Merck
All Photos(1)

Key Documents

398853

Sigma-Aldrich

Lead(II) bromide

99.999% trace metals basis

Synonym(s):

Lead dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbBr2
CAS Number:
Molecular Weight:
367.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
powder

Quality Level

Assay

99.999% trace metals basis

form

powder

reaction suitability

core: lead

impurities

≤15.0 ppm Trace Metal Analysis

bp

892 °C (lit.)

mp

371 °C (lit.)

density

6.66 g/mL at 25 °C (lit.)

SMILES string

Br[PbH2]Br

InChI

1S/2BrH.Pb/h2*1H;/q;;+2/p-2

InChI key

ZASWJUOMEGBQCQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lead(II) bromide is a white crystalline solid that is sparingly soluble in water. It possesses excellent optical properties, including a suitable bandgap for efficient light absorption in the visible spectrum, making it an ideal candidate for photovoltaic applications.

Application

Lead(II) bromide can be used:
  • To prepare an electrolyte for a high performance all-solid-state bromide-ion battery.
  • As a precursor to prepare organic–inorganic hybrid perovskite materials for solar cells and light emitting devices (LEDs).
  • As a starting material to prepare photocatalytic CsPbBr3@SiO2 composites with good water stability.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A N Hamir et al.
Australian veterinary journal, 57(9), 401-406 (1981-09-01)
Eight-month-old dogs maintained on a high-fat-low-calcium diet were administered a mixture of lead chloride, lead bromide and lead sulphate for prolonged periods at 4 different dose levels. Dogs on high levels of leads showed marked weight loss and gastrointestinal symptoms
A O Maslat et al.
Journal of trace elements and electrolytes in health and disease, 3(4), 187-191 (1989-12-01)
The mutagenicity of lead (II) bromide (a combustion product of the gasoline additives lead (IV) tetraethyl and 1,2-dibromoethane) was investigated using various strains of bacteria. Taking prodigiosin (the red pigment) production as a marker, lead (II) bromide was found to
Hongying Duan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(7), 1447-1451 (2004-05-19)
In our experiments, it was observed that adding bromide to Pb2+ solution of N,N'-dimethylformamide (DMF), the highly emissive cluster Pb4Br11(3-) can be formed and the fluorescence intensity of the formed cluster is proportional to the concentration of Pb2+, based on
G Ahmed et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 67(6), 1050-1056 (2009-03-10)
We present the electron momentum densities of PbCl(2) and PbBr(2) using 661.65 keV gamma-rays from 20 Ci (137)Cs source. To supplement our experimental investigations, we also report energy bands, density of states, Mulliken's population and Compton profiles of PbCl(2) and

Articles

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service