Skip to Content
Merck
All Photos(1)

Documents

201014

Sigma-Aldrich

N-Benzylbenzamide

≥98%

Synonym(s):

Benzoic acid benzylamide, N-(Phenylmethyl)benzamide, N-Benzoylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONHCH2C6H5
CAS Number:
Molecular Weight:
211.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

mp

104-106 °C (lit.)

solubility

acetone: 25 mg/mL, clear, colorless

SMILES string

O=C(NCc1ccccc1)c2ccccc2

InChI

1S/C14H13NO/c16-14(13-9-5-2-6-10-13)15-11-12-7-3-1-4-8-12/h1-10H,11H2,(H,15,16)

InChI key

LKQUCICFTHBFAL-UHFFFAOYSA-N

General description

N-Benzylbenzamide inhibits the activity of tyrosinase.

Application

A convenient precursor to α-substituted benzylamines and an indicator for the titration of butyllithium and other lithium bases. For references see Aldrichimica Acta .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manting Chiang et al.
Pharmaceutical research, 37(10), 205-205 (2020-09-30)
Modulation of 5-HT3 receptor in the central nervous system (CNS) is a promising approach for treatment of neuropathic pain. The goal was to evaluate the role of P-glycoprotein (Pgp) in limiting exposure of different parts of the CNS to ondansetron
Yae Eun Chong et al.
Biomedical chromatography : BMC, 33(11), e4653-e4653 (2019-07-20)
Ondansetron, a widely used antiemetic agent, is a P-glycoprotein (P-gp) substrate and therefore expression of P-gp at the blood-brain barrier limits its distribution to the central nervous system (CNS), which was observed to be reversed by coadministration with P-gp inhibitors.
Sung Jin Cho et al.
Bioorganic & medicinal chemistry letters, 16(10), 2682-2684 (2006-03-04)
A series of potent inhibitors of tyrosinase and their structure-activity relationships are described. N-Benzylbenzamide derivatives (1-21) with hydroxyl(s) were synthesized and tested for their tyrosinase inhibitory activity. With this series, compound 15 provided a potent tyrosinase inhibition: it effectively inhibited
Aldrichimica Acta, 11, 20-20 (1978)
Zsanett Dorkó et al.
Talanta, 162, 167-173 (2016-11-14)
A simple and efficient method is presented for assessing molecularly imprinted polymers (MIP) and other sorbents from the point of view of practical applications. The adsorption isotherms of the compounds, which need to be separated or detected in an application

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service