Skip to Content
Merck
All Photos(3)

Documents

108227

Sigma-Aldrich

Benzanilide

98%

Synonym(s):

N-Benzoylaniline, N-Phenylbenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONHC6H5
CAS Number:
Molecular Weight:
197.23
Colour Index Number:
42095
Beilstein:
1102980
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

117 °C/10 mmHg (lit.)

mp

161-163 °C (lit.)

solubility

H2O: insoluble
diethyl ether: slightly soluble

SMILES string

O=C(Nc1ccccc1)c2ccccc2

InChI

1S/C13H11NO/c15-13(11-7-3-1-4-8-11)14-12-9-5-2-6-10-12/h1-10H,(H,14,15)

InChI key

ZVSKZLHKADLHSD-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

General description

Benzanilide undergoes diarylation with aryl triflates or bromides in the presence of a palladium-based catalyst system to form corresponding N-(2,6-diarylbenzoyl)anilines. It is used in manufacture of dyes and perfumes.

Application

Benzanilide was used to study the influence of β-cyclodextrin on photorearrangement of acetanilide, benzanilide and ethyl phenyl carbonate. It was used as amide model compound to study the reaction between the amide and epoxy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dritan Hasa et al.
Pharmaceutics, 12(11) (2020-11-20)
Dodeca-2E,4E,8Z,10E/Z-tetraenoic isobutylamide (tetraene) is the main component of Echinacea angustifolia DC. lipophilic extract, the bioavailability and immunomodulatory effect after oral administration in soft gel capsules in healthy volunteers of which we have already demonstrated. In the present work, we assessed
Wan-Ping Hu et al.
Bioorganic & medicinal chemistry, 16(9), 5295-5302 (2008-03-25)
A series of novel thiobenzanilides is described. These compounds have been previously found to show strong biological activity such as antimycotic and antifungal actions. This is the first demonstration on the mechanism of the anticancer effect of thiobenzanilide agents (4a-c)
Matthew Danish et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(3), 355-359 (2009-01-02)
An analytical method was developed and validated for the quantitative determination of the histone deacetylase inhibitor MS-275 in human plasma. Calibration curves were linear in the concentration range of 1-250 ng/mL. Sample pretreatment involved a liquid-liquid extraction of 0.1 mL
Regioselective arylation of benzanilides with aryl triflates or bromides under palladium catalysis.
Kametani Y, et al.
Tetrahedron Letters, 41(15), 2655-2658 (2000)
Sarah R Dennison et al.
European biophysics journal : EBJ, 41(8), 687-693 (2012-07-12)
The activity of a membrane interactive cis and trans benzanilide against Escherichia coli membrane mimics was investigated using Langmuir monolayers. It was found that in the presence of E. coli lipid mix monolayers, cis-benzanilide induced maximal surface pressure changes of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service