Skip to Content
Merck
All Photos(1)

Key Documents

188956

Sigma-Aldrich

4-Phenylurazole

98%

Synonym(s):

4-Phenyl-1,2,4-triazolidine-3,5-dione, Ph-Ur for e-Y-CLICK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
Beilstein:
163361
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

207-209 °C (lit.)

SMILES string

O=C1NNC(=O)N1c2ccccc2

InChI

1S/C8H7N3O2/c12-7-9-10-8(13)11(7)6-4-2-1-3-5-6/h1-5H,(H,9,12)(H,10,13)

InChI key

GOSUFRDROXZXLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenylurazole undergoes oxidation in the presence of NO2-N2O4 to yield 4-phenyl-1,2,4-trizoline-3,5-dione. It undergoes acetylation reaction with excess acetyl chloride in N,N-dimethylacetamide solution. It polymerizes in the presence of phosgene, terephthaloyl chloride and epichlorohydrin to yield insoluble polymer. It is precursor to the Diels-Alder trapping agent, 4-phenyl-1,2,4-triazoline-3,5-dione. This urazole was recently demonstrated in the traceless, chemoselective labeling of peptides and proteins through electrochemical tyrosine-click (e-Y-CLICK) chemistry.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemische Berichte, 121, 185-185 (1988)
Polycondensation Reaction of 4-(4'-N-1, 8-Naphthalimidophenyl)-1, 2, 4-triazolidine-3, 5-dione with Aliphatic Diacid Chlorides.
Mallakpour S and Rafiee Z.
Iranian Polymer Journal, 13, 225-234 (2004)
Journal of the American Chemical Society, 109, 3730-3730 (1987)
Bulletin of the Chemical Society of Japan, 61, 3915-3915 (1988)
Synthesis of aliphatic polyamides containing 4-phenylurazole linkages.
Mallakpour SE and Sheikholeslami B.
Polymer International, 48(1), 41-46 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service