Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.559 (lit.)
bp
168-169 °C/2 mmHg (lit.)
density
1.159 g/mL at 25 °C (lit.)
SMILES string
OCCc1ccccc1O
InChI
1S/C8H10O2/c9-6-5-7-3-1-2-4-8(7)10/h1-4,9-10H,5-6H2
InChI key
ABFCOJLLBHXNOU-UHFFFAOYSA-N
General description
2-Hydroxyphenethyl alcohol reacts with triethyl or trimethyl aluminium to yield cyclic products and organoaluminim oligomers. Inelastic electron tunnelling spectra of 2-hydroxyphenethyl alcohol adsorbed on thin-film aluminium and magnesium oxide has been investigated.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
An IETS study of the adsorption of 1, 2-, 1, 3-and 1, 4-benzenedimethanol, 2-, 3-and 4-hydroxyphenethyl alcohol, and 2-, 3-and 4-hydroxybenzyl alcohol, on thin-film plasma-grown aluminium and magnesium oxides.
Spectrochimica Acta Part A: Molecular Spectroscopy, 48(7), 939-951 (1992)
Drug metabolism and disposition: the biological fate of chemicals, 12(5), 543-549 (1984-09-01)
A novel metabolite of coumarin was isolated by high pressure liquid chromatography from in vitro incubations with a rat liver 10,000g postmitochondrial supernatant fraction. Mass spectral analysis and co-chromatography with authentic reference compound was utilized to identify the lactone ring-opened
Organic & biomolecular chemistry, 6(16), 2982-2987 (2008-08-09)
We describe the synthesis and NMR spectroscopic analysis of three artificial lignin polymers containing only the beta-O-4 substructure: syringyl-type homopolymer, p-hydroxyphenyl-type homopolymer and guaiacyl/syringyl-type heteropolymer. Using gel permeation chromatography, the weight-average degree of polymerization (DP(w)) of the three polymers was
The reactions of trialkylaluminium with 2-hydroxybenzyl-and 2-hydroxyphenethyl alcohols.
Main Group Metal Chemistry, 21(2), 105-112 (1998)
Food chemistry, 285, 275-281 (2019-02-25)
The exposure of fatty products to environmental light can trigger lipid oxidation in food through a sensitized-photooxidation process, which involves the participation of the species singlet oxygen (O2(1Δg)). Therefore, preservation of food quality represents a subject of great economic interest
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service