Skip to Content
Merck
All Photos(1)

Key Documents

185590

Sigma-Aldrich

Propionitrile

99%

Synonym(s):

Ethyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CN
CAS Number:
Molecular Weight:
55.08
Beilstein:
773680
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.366 (lit.)

bp

97 °C (lit.)

mp

−93 °C (lit.)

solubility

water: soluble 11.9g/100g at 40 °C
water: soluble 29g/100g at 100 °C
DMF: miscible
alcohol: miscible
diethyl ether: miscible

density

0.772 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

CCC#N

InChI

1S/C3H5N/c1-2-3-4/h2H2,1H3

InChI key

FVSKHRXBFJPNKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propionitrile is an experimental duodenal ulcerogen and was found to stimulate gastric acid secretion in the rat.

Application

Propionitrile was used in mesoporous graphitic C3N4 catalyzed cyclotrimerisation of various nitriles into triazine derivatives. It was used in Raman spectroscopic study of complex formation between o-cresol and propionitrile.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Szabo et al.
Research communications in chemical pathology and pharmacology, 16(2), 311-323 (1977-02-01)
Cysteamine and propionitrile, experimental duodenal ulcerogens, stimulated gastric acid secretion in the rat. Gastric acid secretion was measured by two separate methods, the conventional pylorus ligation technique and a non-invasive technique based on the pH dependent liberation of azure A
A Raman spectroscopic study of complex formation between o-cresol and propionitrile.
Girling RB and Shurvell HF.
Vibrational Spectroscopy, 18(1), 77-82 (1998)
D Somjen et al.
Journal of cellular biochemistry, 112(2), 625-632 (2011-01-27)
In cultured human osteoblasts estradiol-17β (E2) modulated DNA synthesis, the specific activity of creatine kinase BB (CK), 12 and 15 lipoxygenase (LO) mRNA expression and formation of 12- and 15-hydroxyeicosatetraenoic acid (HETE). We now investigate the response of human bone
H V O Carswell et al.
American journal of physiology. Heart and circulatory physiology, 287(4), H1501-H1504 (2004-05-25)
The present study employs selective estrogen receptor (ER) agonists to determine whether 17beta-estradiol-induced neuroprotection in global ischemia is receptor mediated and, if so, which subtype of receptor (ERalpha or ERbeta) is predominantly responsible. Halothane-anesthetized female C57Bl/6J mice were ovariectomized, and
Oliver Kaumanns et al.
The Journal of organic chemistry, 74(1), 75-81 (2008-11-27)
The rates of the reactions of the colored para-substituted phenylacetonitrile anions 1a-c and the phenylpropionitrile anions 2a-c with Michael acceptors (3a-u) were determined by UV-vis spectroscopy in DMSO at 20 degrees C. The reactions follow second-order kinetics, and the corresponding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service