Skip to Content
Merck
All Photos(4)

Key Documents

110213

Sigma-Aldrich

Acrylonitrile

≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

Vinyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCN
CAS Number:
Molecular Weight:
53.06
Beilstein:
605310
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

1.83 (vs air)

Quality Level

vapor pressure

86 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

897 °F

contains

35-45 ppm monomethyl ether hydroquinone as inhibitor

expl. lim.

17 %

refractive index

n20/D 1.391 (lit.)

bp

77 °C (lit.)

mp

−83 °C (lit.)

SMILES string

C=CC#N

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2

InChI key

NLHHRLWOUZZQLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acrylonitrile is commonly used as a monomer which belongs to the class of unsaturated nitriles. It serves as a key monomer in the production of various polymers and copolymers with desirable properties. Acrylonitrile is commonly employed in the production of acrylic fibers, which are known for their softness, strength, and excellent thermal and chemical resistance. One of the primary applications of acrylonitrile is in the manufacturing of acrylonitrile-butadiene-styrene (ABS) copolymers. ABS is a versatile and highly durable plastic widely used in automotive components, appliances, electronic enclosures, and consumer goods. The addition of acrylonitrile to the copolymer enhances its heat resistance and impact strength, making ABS a highly sought-after material.

Application

Acrylonitrile can be used as a monomer:
  • In the preparation of the 3D-printed polymer material, Acrylonitrile Butadiene Styrene (ABS) which is a commonly used engineering thermoplastic known for its high strength, durability, and heat resistance. It serves as a suitable substrate for a wide range of applications, including in the medical field, compatible manufacturing processes, injection molding, blow molding, and extrusion.
  • In the copolymerization with lignosulfonate to develop a carbon fiber precursor. This copolymer can serve as a precursor material that undergoes further thermal treatment to produce carbon fibers.
  • To synthesize acrylamide grafted acrylonitrile copolymer membranes, which serve as a support matrix for the immobilization of cellulase enzymes.
Acrylonitrile is used in the manufacture of acrylic fibers, resins (acrylonitrile butadiene-styrene, styrene-acrylonitrile and others) and nitrile rubbers (butadiene-acrylonitrile).

Biochem/physiol Actions

An industrial carcinogen that is a multisite carcinogen in rats and possibly carcinogenic to humans.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Whysner et al.
Regulatory toxicology and pharmacology : RTP, 27(3), 217-239 (1998-08-07)
Acrylonitrile (ACN) exposure is associated with tumors in rat brain, Zymbal gland, and mammary gland. Adducts affecting base pairing were formed in isolated DNA exposed in vitro to the ACN metabolite cyanoethylene oxide (CNEO). DNA from liver, which is not
Acrylonitrile and human cancer--an overview.
D Coggon et al.
Scandinavian journal of work, environment & health, 24 Suppl 2, 81-82 (1998-08-26)
Philip Cole et al.
Regulatory toxicology and pharmacology : RTP, 52(3), 342-351 (2008-10-18)
Several retrospective cohort epidemiology studies evaluated a number of health outcomes in workers exposed to acrylonitrile (AN). The epidemiology studies included in this review have been published since 1970 and were identified through Ovid and MEDLINE retrieval services using search
N A Esmen
Scandinavian journal of work, environment & health, 24 Suppl 2, 63-70 (1998-08-26)
The reconstruction of worker exposures has been the mainstay of modem industrial epidemiologic studies. In most cases, the researchers are faced with the difficult dilemma created by the scarcity of the exposure measurement data vis-à-vis the demands for refined quantification.
A Léonard et al.
Mutation research, 436(3), 263-283 (1999-06-04)
Acrylonitrile (AN) is an important intermediary for the synthesis of a variety of organic products, such as artificial fibres, household articles and resins. Although acute effects are the primary concern for an exposure to AN, potential genotoxic, carcinogenic and teratogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service