Skip to Content
Merck
All Photos(1)

Key Documents

164127

Sigma-Aldrich

2,2′-(4-Methylphenylimino)diethanol

technical grade, 90%

Synonym(s):

2,2′-(p-Tolylimino)diethanol, N,N-Bis(2-hydroxyethyl)-p-toluidine, N-(p-Tolyl)diethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4N(CH2CH2OH)2
CAS Number:
Molecular Weight:
195.26
Beilstein:
2099156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

solid

bp

338-340 °C (lit.)

mp

49-53 °C (lit.)

functional group

amine
hydroxyl

SMILES string

Cc1ccc(cc1)N(CCO)CCO

InChI

1S/C11H17NO2/c1-10-2-4-11(5-3-10)12(6-8-13)7-9-14/h2-5,13-14H,6-9H2,1H3

InChI key

JUVSRZCUMWZBFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-(4-Methylphenylimino)diethanol(2,2′-(p-Tolylimino)diethanol) was used to evaluate the antimicrobial activity of root canal sealers against Enterococcus faecalis and Candida albicans by the agar diffusion test.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prasanna Neelakantan et al.
The Journal of clinical pediatric dentistry, 33(2), 117-122 (2008-01-01)
Sealers with prolonged antimicrobial activity are needed by the fact that microorganisms are commonly recovered from teeth with failed endodontic therapy. The aim of this study was to evaluate the duration of antimicrobial activity of ten root canal sealers (Apexit
K Ikemura et al.
Dental materials journal, 15(2), 132-143 (1996-12-01)
To develop a self-etching primer for ground dentin, 4-acryloxyethyltrimellitic acid (4-AET) was newly synthesized, and the effect of concentrations varying from 0 (control) to 56.5 wt% 4-AET in the water/ HEMA primer on bonding to the dentin was investigated. Bond
S Fujisawa et al.
Journal of dental research, 62(7), 803-805 (1983-07-01)
The interaction between phospholipids and stabilizing lysosome agents such as acetylsalicylic acid (ASA), colchicine (CC), prednisolone (PD), etc., was studied using NMR spectroscopy. The peak intensity of protons arising from these compounds was reduced in liposomes containing lecithin and various
H H Xu et al.
Journal of biomedical materials research, 52(4), 812-818 (2000-10-18)
The strength and toughness of dental core buildup composites in large stress-bearing restorations need to be improved to reduce the incidence of fracture due to stresses from chewing and clenching. The aims of the present study were to develop novel
Ulrich Salz et al.
The journal of adhesive dentistry, 7(1), 7-17 (2005-05-17)
To investigate the influence of the acid-base reaction between acidic monomers and amines on the polymerization behavior of self-etching, self-curing adhesives, determine the effect of the application mode on the shear bond strength and morphology, and elucidate the adhesion performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service