Skip to Content
Merck
All Photos(2)

Key Documents

455709

Sigma-Aldrich

N-tert-Butyldiethanolamine

97%

Synonym(s):

N,N-Bis(2-hydroxyethyl)-tert-butylamine, N-tert-Butyl-2,2′-iminodiethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CN(CH2CH2OH)2
CAS Number:
Molecular Weight:
161.24
Beilstein:
1739942
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.4666 (lit.)

bp

136-139 °C/12 mmHg (lit.)

mp

40-45 °C (lit.)

density

0.983 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CC(C)(C)N(CCO)CCO

InChI

1S/C8H19NO2/c1-8(2,3)9(4-6-10)5-7-11/h10-11H,4-7H2,1-3H3

InChI key

XHJGXOOOMKCJPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiometric detection of N,N'-diethylaminoethanol and lysosomotropic amino alcohols in cation exchange high-performance liquid chromatography systems.
Bazylak G and Nagels LJ.
Analytica Chimica Acta, 472(1), 11-26 (2002)
Dmytro S Nesterov et al.
Chemical communications (Cambridge, England), 50(26), 3431-3434 (2014-01-25)
A complex of copper, possessing an octanuclear structure of high symmetry, has been prepared by a spontaneous self-assembly reaction of Cu(BF4)2 with N-tert-butyldiethanolamine and reveals a paramagnetic ground state.
Frederik Klöwer et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(30), 7413-7422 (2009-06-25)
Three new mu4-bridged Co(II)5 clusters with similar core motifs have been synthesised with the use of N-tert-butyldiethanolamine (tbdeaH2) and pivalic acid (piv): [Co(II)5(mu4-N3)(tbdea)2(mu-piv)4(piv)(CH3CN)2].CH3CN (1), [Co(II)5(mu4-Cl)(Cl)(tbdea)2(mu-piv)4(pivH)2] (2) and [Co(II)5(mu4-N3)(Cl)(tbdea)2(mu-piv)4(pivH)2] (3). Magnetic measurements were performed for all three compounds. It was found
Piotr Król et al.
Colloid and polymer science, 291(4), 1031-1047 (2013-03-26)
Polyurethane elastomers coating were synthesised by using typical diisocyanates, polyether and polyester polyols and HO-tertiary amines or diols as a chain extenders. Mole fractions of structural fragments (
H W Leung et al.
Mutation research, 393(1-2), 7-15 (1997-11-14)
Three alkylalkanolamines, N,N-dimethylethanolamine, N-methyldiethanolamine, and tert-butyldiethanolamine, were evaluated for potential genotoxic activity using the Salmonella/microsome reverse gene mutation test, the CHO/HGPRT forward gene mutation test, a sister chromatid exchange test in cultured CHO cells, and an in vivo peripheral blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service