Skip to Content
Merck
All Photos(1)

Key Documents

161470

Sigma-Aldrich

3-Cyclohexanepropionic acid

99%

Synonym(s):

3-Cyclohexylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CH2CO2H
CAS Number:
Molecular Weight:
156.22
Beilstein:
2042799
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

275.8 °C (lit.)

mp

14-17 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCC1CCCCC1

InChI

1S/C9H16O2/c10-9(11)7-6-8-4-2-1-3-5-8/h8H,1-7H2,(H,10,11)

InChI key

HJZLEGIHUQOJBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3-Cyclohexanepropionic acid can be used:
  • As a starting material in the synthesis of 2-[(1,2,4-triazol-3-yl)thio]acetamide derivatives for in vitro paraoxonase-1 (PON1) studies.
  • To synthesize chemical intermediates such as cyclohexanepropanenitrile , cyclohexanepropanamide , cyclohexanepropanol , and 1-oxaspiro[4.5]decan-2-one .

Biochem/physiol Actions

3-Cyclohexanepropionic acid facilitates oral delivery of cromolyn via permeation across/through the membrane in rats.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

266.0 °F

Flash Point(C)

130 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylenation of aldehydes to 3-propanal, propanol and propanoic acid derivatives
Payne DT, et al.
Scientific reports, 7(1), 1720-1720 (2017)
Remote Allylation of Unactivated C (sp3)-H Bonds Triggered by Photogenerated Amidyl Radicals
Xu B and Tambar UK
ACS Catalysis, 21(7), 947-955 (2019)
Continuous flow synthesis under high-temperature/high-pressure conditions using a resistively heated flow reactor
Adeyemi A, et al.
Organic Process Research & Development, 21(7), 947-955 (2017)
C-H oxygenation at tertiary carbon centers using iodine oxidant
Kiyokawa K, et al.
Chemical Communications (Cambridge, England), 54(55), 7609-7612 (2018)
A Leone-Bay et al.
Pharmaceutical research, 13(2), 222-226 (1996-02-01)
Herein we report the discovery of a group of derivatized alpha-amino acids that increase the oral bioavailability of sodium cromolyn. We prepared three N-acylated alpha-amino acids and used these compounds to demonstrate the oral delivery of cromolyn in an in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service