Skip to Content
Merck
All Photos(2)

Documents

320064

Sigma-Aldrich

Phthalic anhydride

ACS reagent, ≥99%

Synonym(s):

1,2-Benzenedicarboxylic anhydride, 1,3-Isobenzofurandione, Phthalic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4O3
CAS Number:
Molecular Weight:
148.12
Beilstein:
118515
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

5.1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99%
99.0-100.2%

form

flakes

autoignition temp.

1058 °F

expl. lim.

10.4 %

ign. residue

≤0.01%

bp

284 °C (lit.)

mp

131-134 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤5 ppm
heavy metals: ≤5 ppm (by ICP)

SMILES string

O=C1OC(=O)c2ccccc12

InChI

1S/C8H4O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H

InChI key

LGRFSURHDFAFJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phthalic anhydride is an anhydride of phthalic acid and a key intermediate in the production of plasticizers, dyes, polyesters resins, modifiers for rubber, and polymers.

Application

Phthalic anhydride may be used as a reactant in the preparation of N-substituted phthalimides, poly(isothianaphthene) and methyl (S)-2-phthalimido-4-methylthiobutanoate.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

305.6 °F - DIN 51758

Flash Point(C)

152 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New synthetic ?tricks? One-pot preparation of N-substituted phthalimides from azides and phthalic anhydride
Garcia J, et al.
Tetrahedron Letters, 27(5), 639-640 (1986)
Phthalic Acid and Derivatives
Ullmann's Encyclopedia of Industrial Chemistry (2007)
New synthetic routes to poly (isothianaphthene) I. Reaction of phthalic anhydride and phthalide with phosphorus pentasulfide.
van Asselt R, et al.
Synt. Metals, 74(1), 65-70 (1995)
Ceramic foams directly-coated with flame-made V2O5/TiO2 for synthesis of phthalic anhydride
Schimmoeller B, et al.
J. Catal., 243(1), 82-92 (2006)
Methyl (S)-2-phthalimido-4-oxobutanoate
Meffre P, et al.
Organic Syntheses, 76, 123-123 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service