157449
β-Bromostyrene
97%
Synonym(s):
beta-Bromostyrene, 1-Bromo-2-phenylethylene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH=CHBr
CAS Number:
Molecular Weight:
183.05
Beilstein:
2038495
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.607 (lit.)
bp
110-112 °C/20 mmHg (lit.)
mp
7 °C (lit.)
density
1.427 g/mL at 25 °C (lit.)
functional group
bromo
phenyl
storage temp.
2-8°C
SMILES string
Br\C=C\c1ccccc1
InChI
1S/C8H7Br/c9-7-6-8-4-2-1-3-5-8/h1-7H/b7-6+
InChI key
YMOONIIMQBGTDU-VOTSOKGWSA-N
Looking for similar products? Visit Product Comparison Guide
General description
β-Bromostyrene is an α,β-unsaturated aromatic halide. It can be synthesized by catalytic Hunsdiecker reaction (CHR) of cinnamic acid. It is commonly utilized as a precursor for preparing substituted alkenes, corresponding acetylenes and also in the total synthesis of natural compounds and antibiotics.
Application
β-Bromostyrene was used in the one-pot method for the preparation of cinnamonitriles. It was also used in the preparation of pure ethyl Z- and E-α,α-difluoro-4-phenyl-3-butenoate and 1,3-diphenyl-1-butene. It may be used in the synthesis of β-tert-butylstyrene via cross coupling reaction with tert-butylmagnesium chloride in the presence of dichloro[1,1′-bis(diphenylphosphino)ferrocene]nickel(II) catalyst. It may also be used to prepare γ-but-2-enolactone by reacting with nickel carbonyl in the presence of alkynes.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
215.6 °F - closed cup
Flash Point(C)
102 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cross-Coupling of Tertiary Alkyl Grignard Reagents with β-Bromostyrene Catalyzed by Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel (II).
Hayashi T, et al.
Chemistry Letters (Jpn), 9(6), 767-768 (1980)
Enantioselective Preparation of C2-Symmetrical Ferrocenyl Ligands for Asymmetric Catalysis.
Schwink L and Knochel P.
Chemistry (Weinheim An Der Bergstrasse, Germany), 4(5), 950-968 (1998)
The reaction of ?-bromostyrene with nickel carbonyl in the presence of alkynes.
Ryang M, et al.
Journal of Organometallic Chemistry, 46(2), 375-377 (1972)
Practical one-pot conversion of aryl bromides and ?-bromostyrenes into aromatic nitriles and cinnamonitriles.
Ishii G, et al.
Tetrahedron, 69(5), 1462-1469 (2013)
New method for the synthesis of β-bromostyrenes.
Shastin AV, et al.
Russian Chemical Bulletin, 50(8), 1401-1405 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service