Skip to Content
Merck
All Photos(1)

Documents

156833

Sigma-Aldrich

Nonanoyl chloride

96%

Synonym(s):

Pelargonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COCl
CAS Number:
Molecular Weight:
176.68
Beilstein:
1362621
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

108-110 °C/22 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCC(Cl)=O

InChI

1S/C9H17ClO/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3

InChI key

NTQYXUJLILNTFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nonanoyl chloride was used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of:
  • N-nonanoyl piperonylamide
  • caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide
  • n-benzylnonanamide

Other Notes

Remainder 2-Methyloctanoyl chloride

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

210.2 °F

Flash Point(C)

99 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of Acylation and Crosslinking on the Material Properties and Cadmium Ion Adsorption Capacity of Porous Chitosan Beads.
Hsien, Tzu-Yang, and Gregory L. Rorrer
Separation Science and Technology, 30(12), 2455-2475 (1995)
Hypotensive and antinociceptive effects of ether-linked and relatively non-pungent analogues of< i> N</i>-nonanoyl vanillylamide.
Chen IJ , et al.
European Journal of Medicinal Chemistry, 27(3), 187-192 (1992)
Karen C Thomas et al.
The Journal of pharmacology and experimental therapeutics, 321(3), 830-838 (2007-03-03)
Transient receptor potential vanilloid 1 (TRPV1) is a calcium-selective ion channel expressed in human lung cells. We show that activation of the intracellular subpopulation of TRPV1 causes endoplasmic reticulum (ER) stress and cell death in human bronchial epithelial and alveolar
Jun Zhao et al.
Biochemistry, 45(15), 4915-4926 (2006-04-12)
Nociceptive neurons in the peripheral nervous system detect noxious stimuli and report the information to the central nervous system. Most nociceptive neurons express the vanilloid receptor, TRPV1, a nonselective cation channel gated by vanilloid ligands such as capsaicin, the pungent
Yannick Hövelmann et al.
Journal of agricultural and food chemistry, 67(13), 3670-3678 (2019-03-13)
Imidazole alkaloids represent a rather small group of alkaloids and are assumed not to be of significance to the human food chain so far. In this study, novel imidazole alkaloids occurring in tomato products were synthesized and structurally characterized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service