Skip to Content
Merck
All Photos(1)

Key Documents

141615

Sigma-Aldrich

4-Nitroimidazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3N3O2
CAS Number:
Molecular Weight:
113.07
Beilstein:
2815
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

303 °C (dec.) (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1c[nH]cn1

InChI

1S/C3H3N3O2/c7-6(8)3-1-4-2-5-3/h1-2H,(H,4,5)

InChI key

VYDWQPKRHOGLPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitroimidazole is an intermediate during the synthesis of 1-methyl-2,4,5-trinitro imidazole.

Application

4-Nitroimidazole was used in a study to investigate the catalytic efficiency of heterocyclic compounds in the peroxyoxalate chemiluminescence reaction using bis(2,4,6-trichlorophenyl)oxalate as reagent.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Pandurang M Jadhav et al.
Journal of molecular modeling, 19(8), 3027-3033 (2013-04-12)
1-Methyl-2,4,5-trinitro imidazole (MTNI) is a well-known melt cast explosive possessing good thermal stability and impact insensitivity. MTNI has been synthesized from multi-step nitration followed by methylation of imidazole exhibiting low yield. It is desirable to screen the process thermodynamically for
Heterocyclic compounds as catalysts in the peroxyoxalate chemiluminescence reaction of bis (2, 4, 6-trichlorophenyl) oxalate.
Jonsson T and Irgum K.
Analytica Chimica Acta, 361(3), 205-215 (1998)
Javier Giglio et al.
Nuclear medicine and biology, 39(5), 679-686 (2012-04-27)
The evaluation of oxygenation status of solid tumors is an important field of radiopharmaceutical research. With the aim to develop new potential 99mTc-radiopharmaceuticals for imaging hypoxia, we have synthesized two novel isocyanide derivatives of metronidazole, which has demonstrated high affinity
Javier Giglio et al.
Bioorganic & medicinal chemistry letters, 21(1), 394-397 (2010-11-23)
With the aim to develop new potential (99m)Tc-radiopharmaceuticals for imaging hypoxia based on the formation of Tc-nitrido complexes, two novel dithiocarbamate containing metronidazole derivatives (L1 and L2) have been prepared and characterised. The synthesis of L1 and L2 was achieved
David Leitsch et al.
The Journal of antimicrobial chemotherapy, 66(8), 1756-1765 (2011-05-24)
The mechanism of action of, and resistance to, metronidazole in the anaerobic (or micro-aerotolerant) protozoan parasite Giardia lamblia has long been associated with the reduction of ferredoxin (Fd) by the enzyme pyruvate:ferredoxin oxidoreductase (PFOR) and the subsequent activation of metronidazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service