Skip to Content
Merck
All Photos(2)

Key Documents

136824

Sigma-Aldrich

3,3-Dimethyl-2-butanol

98%

Synonym(s):

tert-Butyl methyl carbinol, Pinacoline alcohol, Pinacolyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH(OH)CH3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718948
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.415 (lit.)

bp

119-121 °C (lit.)

mp

4.8 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(O)C(C)(C)C

InChI

1S/C6H14O/c1-5(7)6(2,3)4/h5,7H,1-4H3

InChI key

DFOXKPDFWGNLJU-UHFFFAOYSA-N

General description

3,3-Dimethyl-2-butanol is a potential precursor for prohibited chemical weapons such as soman, a nerve agent. It is a synthetic analog of kairomone.

Application

3,3-Dimethyl-2-butanol (pinacolyl alcohol) can be used as a substrate:
  • To study the oxidation of secondary alcohols to ketones using cyclic microwave heating technique.
  • To prepare aryl ethers by reacting with aryl iodide using 4-pyrrolidinopyridine ligand via Cu-catalyzed Ullmann reaction.
3,3-Dimethyl-2-butanol was used in conversion of ribose- and glucose-binding proteins into receptors for pinacolyl methyl phosphonic acid.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast oxidation of secondary alcohols by the bromate-bromide system using cyclic microwave heating in acidic water
Paakkonen S, et al.
Tetrahedron Letters, 51(51), 6695-6699 (2010)
Chemical ionization mass spectral analysis of pinacolyl alcohol and development of derivatization method using p-tolyl isocyanate.
Murty MRVS, et al.
Analytical Methods : Advancing Methods and Applications, 2(10), 1599-1605 (2010)
3, 3-Dimethyl-1-butanol, a parakairomone component to Aleurodicus dispersus (Hemiptera: Aleyrodidae).
Zheng L-X, et al.
Arthropod-Plant Interactions, 7, 1-7 (2013)
J T James et al.
Journal of applied toxicology : JAT, 7(2), 135-142 (1987-04-01)
Four groups of male and female Sprague-Dawley rats were exposed for 13 weeks to 3,3-dimethyl-2-butanol (PA) at concentrations of 0.00, 0.20, 1.00 or 5.00 mg/l (1 mg/l = 240 ppm). Exposures were for 6 hr per day, 5 days per
J T James et al.
Journal of applied toxicology : JAT, 7(5), 307-312 (1987-10-01)
Sprague-Dawley rats were given 15, 70 and 140 min exposures to 15 mg/l 3,3-dimethyl-2-butanol, pinacolyl alcohol (PA), or 6-hour exposures to 0.2, 1.0 and 5.0 mg/l PA (1 mg/l = 240 ppm). A 50% mortality rate was obtained at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service