Skip to Content
Merck
All Photos(1)

Key Documents

129240

Sigma-Aldrich

2,3-Dimethyl-2-butanol

98%

Synonym(s):

Thexyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC(OH)(CH3)2
CAS Number:
Molecular Weight:
102.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

120-121 °C (lit.)

mp

−14 °C (lit.)

density

0.823 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)C(C)(C)O

InChI

1S/C6H14O/c1-5(2)6(3,4)7/h5,7H,1-4H3

InChI key

IKECULIHBUCAKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dimethyl-2-butanol is a major product of aging[1].

Application

2,3-Dimethyl-2-butanol was used to study the absolute rate co-efficient of the reaction of hydroxyl radical with 2,3-dimethyl-2-butanol[2].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Jiménez et al.
The journal of physical chemistry. A, 109(48), 10903-10909 (2005-12-08)
The absolute rate coefficients for the reactions of hydroxyl radical (OH) with 2-butanol (k(1)), 2-methyl-2-butanol (k(2)), and 2,3-dimethyl-2-butanol (k(3)) were measured as a function of temperature (263-354 K) and pressure (41-193 Torr of He, Ar, and N(2)) by the pulsed
Gulseher Sarah Sirin et al.
The journal of physical chemistry. B, 116(40), 12199-12207 (2012-09-19)
Acetylcholinesterase (AChE) is a crucial enzyme in the cholinergic nervous system that hydrolyzes neurotransmitter acetylcholine (ACh) and terminates synaptic signals. The catalytic serine of AChE can be phosphonylated by soman, one of the most potent nerve agents, and subsequently undergo
Jia Wang et al.
Lab on a chip, 19(14), 2415-2424 (2019-06-13)
Application of microfluidics offers numerous advantages in the field of radiochemistry and could enable dramatic reductions in the cost of producing radiotracers for positron emission tomography (PET). Droplet-based microfluidics, in particular, requires only microgram quantities of expensive precursors and reagents
Jia Wang et al.
Lab on a chip, 17(24), 4342-4355 (2017-11-23)
Despite the increasing importance of positron emission tomography (PET) imaging in research and clinical management of disease, access to myriad new radioactive tracers is severely limited due to their short half-lives (which requires daily production) and the high cost and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service