Skip to Content
Merck
All Photos(1)

Key Documents

132179

Sigma-Aldrich

2,4,6-Trimethylaniline

98%

Synonym(s):

Mesidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2NH2
CAS Number:
Molecular Weight:
135.21
Beilstein:
636559
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.551 (lit.)

bp

108-110 °C/11 mmHg (lit.)
233 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)c(N)c(C)c1

InChI

1S/C9H13N/c1-6-4-7(2)9(10)8(3)5-6/h4-5H,10H2,1-3H3

InChI key

KWVPRPSXBZNOHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Synthesis and crystal structures of N,2,4,6-tetra-methyl-anilinium tri-fluoro-methane-sulfonate and N-iso-propyl-idene-N,2,4,6-tetra-methyl-anilinium tri-fluoro-methane-sulfonate.: This study presents the synthesis and structural analysis of two compounds derived from 2,4,6-Trimethylaniline, contributing to understanding its role in organic synthesis and crystal engineering (Stewart et al., 2024).
  • Analysis of aromatic amines in human urine using comprehensive multi-dimensional gas chromatography-mass spectrometry (GCxGC-MS).: This paper explores advanced analytical techniques for detecting aromatic amines, including 2,4,6-Trimethylaniline, demonstrating its relevance in environmental and biological studies (Lorenzo-Parodi et al., 2024).
  • Efficient Rhodium-Catalyzed Multicomponent Reaction for the Synthesis of Novel Propargylamines.: This research highlights the use of 2,4,6-Trimethylaniline in developing new catalytic methods, showcasing its importance in pharmaceutical intermediate synthesis (Rubio-Pérez et al., 2015).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salma Shirin et al.
Environmental science & technology, 45(24), 10369-10377 (2011-11-05)
Sulfonated anthracenedione dyes are medium priority organic compounds targeted for environmental assessment under the Government of Canada's Chemical Management Plan (CMP). Since organic compounds undergo transformations in environmental matrices, understanding these transformations is critical for a proper assessment of their
Ariadna Garza-Ortiz et al.
Inorganic chemistry, 47(15), 6964-6973 (2008-07-03)
The synthetic, spectroscopic, structural, and biological studies of a bis(arylimino)pyridine Ru(III) chloride compound containing the ligand, 2,6-bis(2,4,6-trimethylphenyliminomethyl)pyridine are reported. The bis(arylimino)pyridine ligand, with three donor nitrogen atoms, was synthesized by condensation of 2,6-pyridinedicarboxaldehyde with 2,4,6-trimethylaniline. The Ru(III) complex, with formula
S K Clark et al.
Molecular immunology, 20(12), 1379-1384 (1983-12-01)
Inhibition of the catalytic activity of horseradish peroxidase by rabbit antisera was measured using alternate enzyme substrates. The same general inhibition curve patterns were obtained regardless of the electron donor or hydroperoxide used. In all cases typical biphasic inhibition patterns
2,4,5- and 2,4,6-Trimethylaniline and their hydrochlorides.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 27, 177-188 (1982-04-01)
L Alvarez-Rodríguez et al.
The Analyst, 122(5), 459-463 (1997-05-01)
The spectrophotometric determination of the carbamate pesticides carbaryl, bendiocarb, carbofuran, methiocarb, promecarb and propoxur is proposed. The pesticides are hydrolyzed in alkaline medium to 1-naphthol or phenolates, which are coupled with diazotized trimethylaniline (TMA) in a sodium dodecyl sulfate micellar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service