Skip to Content
Merck
All Photos(3)

Documents

126632

Sigma-Aldrich

1,2-Dinitrobenzene

97%

Synonym(s):

o-Dinitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NO2)2
CAS Number:
Molecular Weight:
168.11
Beilstein:
642224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

319 °C/773 mmHg (lit.)

mp

114-117 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1[N+]([O-])=O

InChI

1S/C6H4N2O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H

InChI key

IZUKQUVSCNEFMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dinitrobenzene (o-Dinitrobenzene) is one of the nitroaromatic compounds present in urine which has been studied using gas chromatography-mass spectrometry-selected ion monitoring methods†. The electrochemistry of 1,2-dinitrobenzene (o-Dinitrobenzene) is strongly affected by 1,3-diphenylurea.

Application

  • A highly divergent Pictet-Spengler approach for pyrrolo quinoxalines from aryl amine using 1,2-dinitrobenzene as an oxidant: Discusses the utility of 1,2-dinitrobenzene as an oxidant in organic synthesis, relevant for developing novel pharmaceuticals (Pardeshi et al., 2019).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cindy Chan-Leonor et al.
The Journal of organic chemistry, 70(26), 10817-10822 (2005-12-17)
[reaction: see text] The electrochemistry of 1,2-dinitrobenzene (1,2-DNB), 1,3-dinitrobenzene (1,3-DNB), and 1,4-dinitrobenzene (1,4-DNB) is strongly affected by the presence of 1,3-diphenylurea. In DMF, the second reduction potential of all three DNBs shifts substantially positive in the presence of the urea
H Tamaki et al.
Journal of bacteriology, 181(9), 2958-2962 (1999-04-28)
Glutathione S-transferases (GSTs) Y-1 and Y-2 from the yeast Issatchenkia orientalis were purified by passage through a glutathione-agarose column, and the cDNA for GST Y-1 was cloned and sequenced. The deduced amino acid sequence consisted of 188 residues with a
J V Barbo et al.
Journal of immunology (Baltimore, Md. : 1950), 154(4), 1551-1559 (1995-02-15)
A population of CD8+ T cells from dinitrobenzene sulfonate-primed mice produce soluble effector molecules that down-regulate the magnitude of dinitrophenol-specific contact hypersensitivity reactions. These soluble molecules express the binding specificity and serologic determinants of alpha/beta TCR. To examine the requirement
Tomoko Andoh et al.
FEBS letters, 525(1-3), 131-134 (2002-08-07)
In Saccharomyces cerevisiae, the overexpression of ROD1 confers resistance to o-dinitrobenzene (o-DNB), a representative of target drugs of glutathione S-transferase. The roles of Rod1 in drug resistance have remained to be determined. We isolated the rog3 mutation as a suppressor
P A Cossum et al.
Drug metabolism and disposition: the biological fate of chemicals, 13(6), 664-668 (1985-11-01)
The metabolism of radiolabeled dinitrobenzene (DNB) isomers was compared in hepatocytes and hepatic subcellular fractions isolated from male Fischer-344 rats. Under aerobic conditions, reduction was the major metabolic pathway for m- and p- DNB in hepatocytes with m- and p-nitroaniline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service