Skip to Content
Merck
All Photos(2)

Documents

45966

Supelco

1,3-Dinitrobenzene

analytical standard

Synonym(s):

1,3-DNB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NO2)2
CAS Number:
Molecular Weight:
168.11
Beilstein:
1105654
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

297 °C (lit.)

mp

84-86 °C (lit.)

density

1.368 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

[O-][N+](=O)c1cccc(c1)[N+]([O-])=O

InChI

1S/C6H4N2O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H

InChI key

WDCYWAQPCXBPJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dinitrobenzene is grouped under the family of nitroaromatic compounds, which find application in the production of pesticides, explosives, polymers, etc. They can pose a potential risk to the ecosystem, owing to their toxic nature.

Application

1,3-Dinitrobenzene has been used as an analytical standard for the quantification of the analyte in organic explosives investigated during the forensic analysis by liquid chromatography-atmospheric pressure chemical ionization mass spectrometry (LC-APCI-MS). It may be used as an analytical standard for the determination of the analyte in aqueous samples, environmental samples and human biological samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Cassada et al.
Journal of chromatography. A, 844(1-2), 87-95 (1999-07-10)
Recent improvements in the LC-MS interface have increased the sensitivity and selectivity of this instrument in the analysis of polar and thermally-labile aqueous constituents. Determination of RDX, nitroso-RDX metabolites, and other munitions was enhanced using LC-MS with solid-phase extraction, 15N3-RDX
Application of DLLME based on the solidification of floating organic droplets for the determination of dinitrobenzenes in aqueous samples
Wu Y, et al.
Chromatographia, 72(7-8), 695-699 (2010)
S Jönsson et al.
Journal of chromatography. A, 1164(1-2), 65-73 (2007-08-04)
A solid-phase microextraction (SPME) method using gas chromatography-electron-capture negative ionisation mass spectrometry (GC-ECNI-MS) and isotope dilution quantification for the analysis of nitroaromatic compounds in complex, water based samples has been optimised. For ionisation, ECNI was the most sensitive and selective
M E Walsh
Talanta, 54(3), 427-438 (2008-10-31)
Hazardous waste site characterization, forensic investigations, and land mine detection are scenarios where soils may be collected and analyzed for traces of nitroaromatic, nitramine, and nitrate ester explosives. These thermally labile analytes are traditionally determined by high-performance liquid chromatography (HPLC);
Romain Tachon et al.
Journal of chromatography. A, 1154(1-2), 174-181 (2007-04-25)
A new LC/MS method was developed for the analysis of sixteen different analytes including the most common organic explosives encountered in forensic investigations. The separation was achieved using a porous graphitic carbon (PGC) column with a binary gradient elution. Molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service