Skip to Content
Merck
All Photos(1)

Key Documents

126349

Sigma-Aldrich

(3,4-Dimethoxyphenyl)acetonitrile

98%

Synonym(s):

Homoveratronitrile, (3,4-Dimethoxyphenyl)acetonitrile, 3,4-Dimethoxybenzylcyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CN
CAS Number:
Molecular Weight:
177.20
Beilstein:
1956100
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

171-178 °C/10 mmHg (lit.)

mp

62-63.5 °C (lit.)

fluorescence

λex 430 nm; λem 511 nm (pH 7.1)

functional group

nitrile

SMILES string

COc1ccc(CC#N)cc1OC

InChI

1S/C10H11NO2/c1-12-9-4-3-8(5-6-11)7-10(9)13-2/h3-4,7H,5H2,1-2H3

InChI key

ASLSUMISAQDOOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(3,4-Dimethoxyphenyl)acetonitrile has been used in the synthesis of (Z)-3-(benzo[b]thiophen-2-yl)-2-(3,4-dimethoxyphenyl)acrylonitrile. It has also been used in the preparation of modified diterpene (±) nimbidiol which relies on a sulfenium ion promoted polyene cyclization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfenium ion promoted polyene cyclizations in natural product synthesis: An efficient biomimetic-like synthesis of (?) nimbidiol.
Harring SR and Livinghouse T.
Tetrahedron Letters, 30(12), 1499-1502 (1989)
Vijayakumar N Sonar et al.
Acta crystallographica. Section C, Crystal structure communications, 63(Pt 12), o743-o745 (2007-12-07)
The title compounds, C20H17NO3S, (I), and C19H15NO2S, (II), were prepared by the reaction of benzo[b]thiophene-2-carbaldehyde with (3,4,5-trimethoxyphenyl)acetonitrile and (3,4-dimethoxyphenyl)acetonitrile, respectively, in the presence of methanolic potassium hydroxide. In (I), the C=C bond linking the benzo[b]thiophene and the 3,4,5-trimethoxyphenyl units has
[Toxicity of 3,4-dimethoxyphenylacetonitrile].
N S Grodetskaia et al.
Gigiena truda i professional'nye zabolevaniia, (2)(2), 55-55 (1982-02-01)
Abdullah Mohamed Asiri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 82(1), 444-455 (2011-08-23)
This work presents the synthesis and characterization of a novel compound, 3-(4-methoxy-phenyl)-2-(4-nitro-phenyl)-acrylonitrile (abbreviated as 3-(4MP)-2-(4-NP)-AN, C(16)H(12)N(2)O(3)). The spectroscopic properties of the compound were examined by FT-IR, UV-vis and NMR ((1)H and (13)C) techniques. FT-IR spectrum in solid state was observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service