Skip to Content
Merck
All Photos(1)

Key Documents

A78608

Sigma-Aldrich

2-Aminopyrimidine

97%

Synonym(s):

2-Pyrimidinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3
CAS Number:
Molecular Weight:
95.10
Beilstein:
107014
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

122-126 °C (lit.)

SMILES string

Nc1ncccn1

InChI

1S/C4H5N3/c5-4-6-2-1-3-7-4/h1-3H,(H2,5,6,7)

InChI key

LJXQPZWIHJMPQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kamaljit Singh et al.
European journal of medicinal chemistry, 52, 82-97 (2012-03-31)
2-Aminopyrimidine based 4-aminoquinolines were synthesized using an efficacious protocol. Some of the compounds showed in vitro anti-plasmodial activity against drug-sensitive CQ(S) (3D7) and drug-resistant CQ(R) (K1) strains of Plasmodium falciparum in the nM range. In particular, 5-isopropyloxycarbonyl-6-methyl-4-(2-nitrophenyl)-2-[(7-chloroquinolin-4-ylamino)butylamino] pyrimidine depicted the lowest
Ye-Xiang Xie et al.
The Journal of organic chemistry, 71(21), 8324-8327 (2006-10-10)
Efficient and solvent-free copper-catalyzed N-arylations of imidazoles with aryl and heteroaryl halides have been demonstrated. In the presence of CuBr, 2-aminopyrimidine-4,6-diol, and TBAF (n-Bu4NF), a variety of imidazoles underwent the N-arylation reaction with aryl and heteroaryl halides smoothly in moderate
Nicholas R Perl et al.
Journal of the American Chemical Society, 132(6), 1802-1803 (2010-01-26)
A convergent synthesis of (-)-crambidine is reported. The sequence capitalizes on two novel key transformations, including a [4+2] annulation of thioimidates with vinyl carbodiimides and an alkyne hydroamination employing 2-aminopyrimidine nucleophiles.
D S Ermolat'ev et al.
Molecular diversity, 15(2), 491-496 (2010-08-27)
An efficient microwave-assisted one-pot two-step protocol was developed for the construction of disubstituted 2-amino-1H-imidazoles. This process involves the sequential formation of 2,3-dihydro-2-hydroxyimidazo[1,2-a]pyrimidinium salts from readily available 2-aminopyrimidines and α-bromoketones, followed by cleavage of the pyrimidine ring with hydrazine.
Rogier A Smits et al.
Drug discovery today, 14(15-16), 745-753 (2009-05-30)
The search for new and potent histamine H4 receptor ligands is leading to a steadily increasing number of scientific publications and patent applications. Several interesting and structurally diverse compounds have been found, but fierce IP competition for a preferred 2-aminopyrimidine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service