Skip to Content
Merck
All Photos(1)

Key Documents

109428

Sigma-Aldrich

4-Hydroxyantipyrine

99%

Synonym(s):

4-Hydroxy-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, NSC 174055

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

184-186 °C (lit.)

functional group

ketone

SMILES string

CN1N(c2ccccc2)C(=O)C(O)=C1C

InChI

1S/C11H12N2O2/c1-8-10(14)11(15)13(12(8)2)9-6-4-3-5-7-9/h3-7,14H,1-2H3

InChI key

SKVPTPMWXJSBTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxyantipyrine is formed during oxidative deamination of aminopyrine. It is a metabolite of antipyrine.

Application

4-Hydroxyantipyrine was used to study the relationships between the metabolism of antipyrine, hexobarbitone and theophylline in man. It was used in a study on flow injection analysis system for the characterisation of pharmaceutical compounds via combination of diode array UV, 1H NMR, FT-IR spectroscopy and time-of-flight mass spectrometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R P Shrewsbury et al.
Research communications in chemical pathology and pharmacology, 67(3), 411-414 (1990-03-01)
Two reports of antipyrine disposition in rats after hemodilution with 20, 40, or 80 ml/kg of Fluosol-DA are evaluated to determine if the extent of hemodilution influenced cytochrome P-450 mediated antipyrine metabolism. Alterations in antipyrine clearance (Cl) and 3-hydroxymethylantipyrine (3OHME)
J H Schellens et al.
British journal of clinical pharmacology, 26(4), 373-384 (1988-10-01)
1. Three model substrates for the characterization of drug oxidation activity, antipyrine (AP), hexobarbitone (HB) and theophylline (TH), were administered to 26 healthy volunteers on two different occasions: in the first experiment a combination of AP (250 mg) and HB
V L Lanchote et al.
Therapeutic drug monitoring, 19(6), 705-710 (1998-01-08)
A method has been developed for the simultaneous determination of antipyrine and its three major metabolites in plasma of patients with renal failure. Plasma samples (500 microl) were hydrolyzed with beta-glucuronidase/aryl sulphatase. The compounds, after addition of sodium chloride, were
R P Shrewsbury et al.
Research communications in chemical pathology and pharmacology, 64(3), 455-462 (1989-06-01)
Antipyrine metabolism was determined after hemodilution with 40 ml/kg of Fluosol in conscious, unrestrained female and male rats. Rats received an intravenous antipyrine dose (20 mg/kg) 24, 48, or 72 hours after hemodilution and the pharmacokinetic parameters were compared to
C A Van Geel et al.
Mechanisms of ageing and development, 53(2), 169-177 (1990-04-09)
The influence of ageing on the metabolism of antipyrine by different forms of cytochrome P-450 was studied in vitro, by measuring the formation of antipyrine metabolites by microsomes isolated from untreated rats, which were grouped into 5 different ages. Km

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service