06720
2-Aminoethyl hydrogen sulfate
≥98.0% (T)
Synonym(s):
Sulfuric acid mono 2-aminoethylester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98.0% (T)
mp
277 °C (dec.) (lit.)
functional group
amine
SMILES string
NCCOS(O)(=O)=O
InChI
1S/C2H7NO4S/c3-1-2-7-8(4,5)6/h1-3H2,(H,4,5,6)
InChI key
WSYUEVRAMDSJKL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Psychopharmacology, 109(1-2), 191-197 (1992-01-01)
Long-term effects of chronic treatment with a GABA-T (GABA-transaminase) inhibitor, ethanolamine O-sulphate (EOS) (200 mg/kg/day for the postnatal days 3-21) on the binding parameters of GABAA receptors, hypothalamic monoamines and subsequent behavior were studied in Wistar rats. At the age
Brain research, 452(1-2), 203-211 (1988-06-14)
Locomotor activity in the rat was studied after infusion of GABAergic and enkephalinergic agonists into the nucleus basalis magnocellularis (NBM) of the forebrain. The experiments were designed to find out whether pharmacological blockade of cholinergic neurons in the NBM had
British journal of pharmacology, 122(3), 539-545 (1997-11-14)
1. The effects of 2, 8 and 21 day oral treatment with the specific gamma-aminobutyric acid transaminase (GABA-T) inhibitors gamma-vinyl GABA (GVG) and ethanolamine O-sulphate (EOS) on brain GABA levels, GABA-T activity, and basal and stimulated GABA release from rat
Psychopharmacology, 89(2), 155-161 (1986-01-01)
Effects of chlordiazepoxide (CDP) and ethanolamine-O-sulphate (EOS) alone and in combination were tested on the acquisition and performance of continuous reinforcement - time out (CR-TO) and variable interval reinforcement - time out (VI-TO) operant discriminations in rats. CDP disrupted acquisition
GABA-mimetic compounds block haloperidol-induced hyperprolactinemia in rats.
Advances in biochemical psychopharmacology, 42, 139-144 (1986-01-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service