Skip to Content
Merck
All Photos(1)

Documents

27980

Sigma-Aldrich

Crotonaldehyde, mixture of cis and trans

ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)

Synonym(s):

2-Butenal, mixture of cis and trans

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCHO
CAS Number:
Molecular Weight:
70.09
Beilstein:
1209254
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.5% (GC)

form

liquid

contains

~0.1% 2,6-di-tert-butyl-4-methylphenol as stabilizer
~1% H2O as stabilizer

refractive index

n20/D 1.437

bp

101-102 °C (lit.)

density

0.853 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C=C\C=O

InChI

1S/C4H6O/c1-2-3-4-5/h2-4H,1H3/b3-2+

InChI key

MLUCVPSAIODCQM-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crotonaldehyde(2-Butenal) is an α, β-unsaturated aldehyde and extremely reactive compound. It serves as a chemical building block in the production of solvents, sorbates, and, to a lower extent, pharmaceuticals and aroma compounds. Additionally, it can be used as an alcohol denaturant, in leather tanning and in the manufacturing of rubber accelerators.

Application

Crotonaldehyde is used:

  • In the manufacturing of sorbic acid that are widely employed as food preservatives to avoid the food spoilage by microorganisms.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

F L Chung et al.
Cancer research, 44(3), 990-995 (1984-03-01)
Acrolein reacted with deoxyguanosine at pH 7 and 37 degrees to give three major products, Adducts 1 to 3, which were separated by high-performance liquid chromatography. They were identified by their ultraviolet, mass, and nuclear magnetic resonance spectra, by the
Eşref Demir et al.
Mutation research, 726(2), 98-103 (2011-07-19)
Lipid-peroxidation products are formed by the thermal treatment of foodstuffs, as well as by endogenous processes. In addition, they are also common environmental pollutants originating from many different sources. Since conflicting data exist on their possible risk for humans, we
Qian-Yuan Wu et al.
Water research, 44(12), 3625-3634 (2010-05-25)
Recent studies have reported that chlorination increased the antiestrogenic activity of wastewater, suggesting that disinfection by-products (DBPs) formed during chlorination is a potential and important source of endocrine-disruptor. However, antiestrogenic DBPs have not been identified. In this study, the antiestrogenic
Organocatalytic asymmetric inverse-electron-demand Diels-Alder reaction of electron-deficient dienes and crotonaldehyde.
Jun-Long Li et al.
Angewandte Chemie (International ed. in English), 49(36), 6418-6420 (2010-07-31)
Seung Eun Lee et al.
Toxicology letters, 201(3), 240-248 (2011-01-18)
Crotonaldehyde, a highly reactive α, β-unsaturated aldehyde, is a ubiquitous environmental pollutant and a product of endogenous lipid peroxidation. It is also a major component of cigarette smoke and is present in many foods and beverages, and has also been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service