Merck
  • Inhibition of delta-secretase improves cognitive functions in mouse models of Alzheimer's disease.
Inhibition of delta-secretase improves cognitive functions in mouse models of Alzheimer's disease.
Nature communications (2017-03-28)
Zhentao Zhang, Obiamaka Obianyo, Elfriede Dall, Yuhong Du, Haian Fu, Xia Liu, Seong Su Kang, Mingke Song, Shan-Ping Yu, Chiara Cabrele, Mario Schubert, Xiaoguang Li, Jian-Zhi Wang, Hans Brandstetter, Keqiang Ye
ABSTRACT
δ-secretase, also known as asparagine endopeptidase (AEP) or legumain, is a lysosomal cysteine protease that cleaves both amyloid precursor protein (APP) and tau, mediating the amyloid-β and tau pathology in Alzheimer's disease (AD). Here we report the therapeutic effect of an orally bioactive and brain permeable δ-secretase inhibitor in mouse models of AD. We performed a high-throughput screen and identified a non-toxic and selective δ-secretase inhibitor, termed compound 11, that specifically blocks δ-secretase but not other related cysteine proteases. Co-crystal structure analysis revealed a dual active site-directed and allosteric inhibition mode of this compound class. Chronic treatment of tau P301S and 5XFAD transgenic mice with this inhibitor reduces tau and APP cleavage, ameliorates synapse loss and augments long-term potentiation, resulting in protection of memory. Therefore, these findings demonstrate that this δ-secretase inhibitor may be an effective clinical therapeutic agent towards AD.
Sigma-Aldrich
delta-Secretase inhibitor, CP11, ≥95% (HPLC)
Sigma-Aldrich
Anti-GFP, N-terminal antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
GSK626616, ≥98% (HPLC)
Anti-GST HRP Conjugate, Cytiva RPN1236, pack of 75 μL