Skip to Content
Merck

SML3355

GSK626616

≥98% (HPLC)

Synonym(s):

(5Z)-2-(2,6-Dichloroanilino)-5-(quinoxalin-6-ylmethylidene)-1,3-thiazolone, (5Z)-2-[(2,6-Dichlorophenyl)amino]-5-(6-quinoxalinylmethylene)-4(5H)-thiazolone, (5Z)-5-(6-Quinoxalinylmethylidene)-2-[(2,6-dichlorophenyl)amino]-1,3-thiazol-4(5H)-one, (Z)-2-(2,6-Dichlorophenylamino)-5-(quinoxalin-6-ylmethylene)thiazol-4(5H)-one, GSK 626616, GSK-626616

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H10Cl2N4OS
CAS Number:
Molecular Weight:
401.27
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC(C=CC=C1Cl)=C1NC2=NC(/C(S2)=C/C3=CC4=C(N=CC=N4)C=C3)=O

InChI

1S/C18H10Cl2N4OS/c19-11-2-1-3-12(20)16(11)23-18-24-17(25)15(26-18)9-10-4-5-13-14(8-10)22-7-6-21-13/h1-9H,(H,23,24,25)/b15-9-

InChI key

RJPNRXFBYZVRIB-DHDCSXOGSA-N

Biochem/physiol Actions

GSK626616 is an orally available, ATP-competitive, potent and selective dual-specificity tyrosine phosphorylation-regulated kinase DYRK inhibitor (DYRK3 IC50 = 0.7 nM; similar potency against DYRK1A and DYRK2) with 20-fold selectivity over the closed related casein kinase 2. GSK626616 enhances Epo-stimulated increase of human marrow erythroid colony formation units (CFU-E), without affecting megakaryocyte colony growth or CFU-GM formation. GSK626616 enhances the percentage and number of Ter119+/CD71+ erythroid progenitors derived from kit+ mouse marrow in the presence of SCF and Epo in cultures, and exhibits therapeutic efficacy in a murine anemia model in vivo (0.03 mg/kg/d i.p.)
Orally available, ATP-competitive, potent and selective dual-specificity tyrosine phosphorylation-regulated kinase DYRK1A/DYRK2/DYRK3 inhibitor in vitro and in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library