Skip to Content
Merck
All Photos(6)

Key Documents

G1544

Sigma-Aldrich

Anti-GFP, N-terminal antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Green Fluorescent Protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 1.0-2.5 μg using GFP fusion protein from transfected mammalian cell lysates
western blot: 0.25-0.5 μg/mL using 3-6 ng of purified GFP fusion proteins expressed in mammalian cell extracts

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Green fluorescent protein (GFP) chromophore contains hexapeptide fragment with Phe64-Ser-Tyr-Gly-Val-Gln69 aminoacid sequence. GFP structure contains a 11-stranded β-barrel and a central helix and corresponds to a molecular weight of 26 kDa.

Specificity

Specific for GFP fusion proteins.

Immunogen

synthetic peptide corresponding amino acids 3-17 of GFP.

Application

Anti-GFP, N-terminal antibody produced in rabbit has been used:
  • in immunoblotting
  • in the histochemistry and immunohistochemistry
  • in the immunoprecipitation


Biochem/physiol Actions

Green fluorescent protein (GFP) has photochemical and diagnostic applications. It is also used widely in protein dynamics studies. The polyclonal anti-GFP, N-terminal rabbit antibody reacts specifically with GFP fusion proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notch signaling controls the differentiation of transporting epithelia and multiciliated cells in the zebrafish pronephros
Liu Y, et al.
Development, 134(6), 1111-1122 (2007)
Green fluorescent protein: a perspective
Remington SJ
Protein Science, 20(9), 1509-1519 (2011)
Green fluorescent protein (GFP): applications, structure, and related photophysical behavior
Zimmer M
Chemical Reviews, 102(3), 759-782 (2002)
Pathogenic mechanism of recurrent mutations of SCN8A in epileptic encephalopathy
Wagnon JL, et al.
Annals of Clinical and Translational Neurology, 3(2), 114-123 (2016)
A phosphorylation-wide sncRNA screen reveals Protein Functional Effector sncRNAs (pfeRNAs) in human lung somatic cells
Gable T, et al.
Cancer letters, 396(6), 85-93 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service