Merck
  • Endocannabinoid and ceramide levels are altered in patients with colorectal cancer.
Endocannabinoid and ceramide levels are altered in patients with colorectal cancer.
Oncology reports (2015-05-16)
Ling Chen, Huixia Chen, Yanting Li, Lei Li, Yan Qiu, Jie Ren
ABSTRACT
Endocannabinoids and ceramides have demonstrated growth inhibition, cell death induction and pro-apoptotic activity in cancer research. In the present study, we describe the profiles of two major endocannabinoids, ceramides, free fatty acids and relevant metabolic enzymes in 47 pairs of human colorectal cancer tissues and adjacent non-tumor tissues. Among them, anandamide (AEA) and its metabolite, arachidonic acid (AA), were markedly upregulated in cancer tissues particularly in those with lymphatic metastasis. The levels of C16 and C24 ceramides were significantly elevated in the colorectal tumor tissues, while levels of C18 and C20 ceramides showed opposite trends. Levels of two enzymes participating in the biosynthesis and degradation of AEA, N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D (NPLD) and fatty acid amide hydrolase (FAAH), together with the most abundant ceramide synthases (CerS1, CerS2, CerS5 and CerS6) in the colon were also determined. Quantitative-PCR analysis indicated that the mRNA levels of these enzymes were overexpressed in the tumor tissues. The activities of NPLD and FAAH were also upregulated. In addition, both gene and protein expression levels of cannabinoid receptor 1 (CB1) were elevated but not of CB2. Elevation of AEA and alteration of ceramides (C16, C24, C18, C20) may qualify as potential endogenous biomarkers and novel drug targets for colorectal cancer.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Methanol, NMR reference standard
Avanti
C20 Ceramide (d17:1/20:0), Avanti Research - A Croda Brand 860648P, powder
Avanti
C24 Ceramide (d17:1/24:0), Avanti Research - A Croda Brand 860649P, powder
Avanti
C24:1 Ceramide (d17:1/24:1(15Z)), Avanti Research - A Croda Brand 860650P, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Avanti
C24 Ceramide-d7 (d18:1-d7/24:0), Avanti Research - A Croda Brand 860678P, powder
Avanti
C12 Ceramide (d18:1/12:0), Avanti Research - A Croda Brand
Avanti
C20 Ceramide (d18:1/20:0), Avanti Research - A Croda Brand 860520P, powder