Merck
Single step transformation of sulphur to Li2S2/Li2S in Li-S batteries.
Scientific reports (2015-07-16)
M Helen, M Anji Reddy, Thomas Diemant, Ute Golla-Schindler, R Jürgen Behm, Ute Kaiser, Maximilian Fichtner
ABSTRACT
Lithium-sulphur batteries have generated tremendous research interest due to their high theoretical energy density and potential cost-effectiveness. The commercial realization of Li-S batteries is still hampered by reduced cycle life associated with the formation of electrolyte soluble higher-order polysulphide (Li2Sx, x = 4-8) intermediates, leading to capacity fading, self-discharge, and a multistep voltage profile. Herein, we have realized a practical approach towards a direct transformation of sulphur to Li2S2/Li2S in lithium-sulphur batteries by alteration of the reaction pathway. A coconut shell derived ultramicroporous carbon-sulphur composite cathode has been used as reaction directing template for the sulphur. The lithiation/delithiation and capacity fading mechanism of microporous carbon confined sulphur composite was revealed by analyzing the subsurface using X-ray photoelectron spectroscopy. No higher-order polysulphides were detected in the electrolyte, on the surface, and in the subsurface of the cathode composite. The altered reaction pathway is reflected by a single-step profile in the discharge/charge of a lithium-sulphur cell.
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Lithium, foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%
Lithium, foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%
Sigma-Aldrich
Lithium, foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%
Sigma-Aldrich
Dimethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Dimethyl carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Lithium-7Li, ≥99.8 atom % 7Li, ≥99.8% (CP)