Skip to Content
Merck
All Photos(2)

Documents

GF40266682

Sigma-Aldrich

Lithium

foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%

Synonym(s):

Lithium, LI000220

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li
CAS Number:
Molecular Weight:
6.94
MDL number:
UNSPSC Code:
12141803
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 402-666-82

resistivity

9.446 μΩ-cm, 20°C

size × thickness

38x500 mm × 0.20 mm

bp

1342 °C (lit.)

mp

180 °C (lit.)

density

0.534 g/mL at 25 °C (lit.)

SMILES string

[Li]

InChI

1S/Li

InChI key

WHXSMMKQMYFTQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Application

  • A retrospective on lithium-ion batteries: This article reviews the contributions in the development of lithium-ion batteries, discussing inspirational insights to guide future breakthroughs in battery technology (J Xie, YC Lu, 2020).
  • Guidelines and trends for next-generation rechargeable lithium and lithium-ion batteries: This review discusses the limitations of commercial lithium-ion batteries and explores the directions for next-generation batteries (F Wu, J Maier, Y Yu, 2020).
  • Prospects for lithium-ion batteries and beyond—a 2030 vision: This article explores the future of lithium-ion batteries and discusses current strategies to improve these and next-generation battery technologies (CP Grey, DS Hall, 2020).
  • Lithium brines: A global perspective: This study offers a global perspective on lithium brines, discussing the critical and technologically important roles of lithium, which is in increasing demand (LA Munk, SA Hynek, DC Bradley, D Boutt, K Labay, 2016).

Legal Information

Product of Goodfellow

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yingying Lu et al.
Nature materials, 13(10), 961-969 (2014-08-12)
Rechargeable lithium, sodium and aluminium metal-based batteries are among the most versatile platforms for high-energy, cost-effective electrochemical energy storage. Non-uniform metal deposition and dendrite formation on the negative electrode during repeated cycles of charge and discharge are major hurdles to
Rasmus W Licht
CNS neuroscience & therapeutics, 18(3), 219-226 (2011-11-11)
Still after more than 50 years, lithium is a major treatment of bipolar disorder, even though it has not been promoted by the pharmaceutical industry over the last decades. In recent years the evidence base on lithium for bipolar disorder
Annemiek Dols et al.
International clinical psychopharmacology, 28(6), 287-296 (2013-07-23)
Side effects are among the most frequent reasons preventing patients from taking their medication. Although the management of side effects is an important issue in clinical practice, particularly in patients with physical comorbidities, research on clinical management of side effects
Mustafa Khasraw et al.
BMC medicine, 10, 131-131 (2012-11-06)
Neurocognitive impairment is being increasingly recognized as an important issue in patients with cancer who develop cognitive difficulties either as part of direct or indirect involvement of the nervous system or as a consequence of either chemotherapy-related or radiotherapy-related complications.
Jiehua Liu et al.
Advanced materials (Deerfield Beach, Fla.), 24(30), 4097-4111 (2012-04-17)
Two dimensional nanoarchitectures are of great interest in lithium storage for energy-storage devices, in particular lithium-ion batteries, due to its shortened paths for fast lithium ion diffusion and large exposed surface offering more lithium-insertion channels. Their competitive lithium-storage features provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service