Merck
Immune Regulation of Interleukin-27 in Malignant Pleural Effusion.
Chinese medical journal (2015-07-15)
Shi Li, Wen-Jie You, Jian-Chu Zhang, Qiong Zhou, Huan-Zhong Shi
ABSTRACT
Interleukin (IL)-27 has been reported to have anti-proliferate and anti-angiogenic activities on cancer cells. However, the involvement of IL-27 in malignant pleural effusion (MPE) remains unknown. Thus, in this research, we compared the immune functions of IL-27, interferon (IFN)-γ, and IL-17 on lung cancer cells and revealed the regulatory mechanism of IL-27 in MPE. The distribution of IL-27 in both MPE and blood was evaluated by enzyme-linked immunosorbent assay and flow cytometry. The expressions of cytokine receptors and the levels of the phosphorylated signal transducer and activator of transcription (STAT) signalings were detected by flow cytometry. As well as the effects of proliferation, apoptosis, migration, and adherent activity of IL-27, IFN-γ, and IL-17 on lung cancer cells were also explored. The expression of IL-27 was increased in MPE when compared with blood (147.3 ± 25.1 pg/ml vs. 100.3 ± 13.9 pg/ml, P = 0.04). IL-27 was noted to suppress both proliferation (18.33 ± 0.21 vs. 27.77 ± 0.88, P = 0.0005) and migration (1.82 ± 0.44 vs. 3.13 ± 0.07, P = 0.04) of A549 cells, but obviously promoted apoptosis of A549 cells (9.47 ± 1.14 vs. 4.96 ± 0.17, P = 0.02) by activating STAT1 signaling. Interestingly, IL-27 played totally opposite effects on A549 cells by activating STAT3 pathway. Moreover, IL-27 exerted different intercellular adherent activities of A549 cells to pleural mesothelial cell monolayer by activating different STAT signalings. IL-27 might exert an important immune regulation on lung cancer cells in human pleural malignant environment.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
IL-17D human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.