Skip to Content
Merck
All Photos(1)

Key Documents

SRP3082

Sigma-Aldrich

IL-17D human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

IL-27

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

40.5 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL17D(53342)

General description

IL-17D is a disulfide-linked homodimer of two 185 amino acid polypeptide chains. It belongs to the IL-17 family of structurally-related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17D has the ability to stimulate the production of IL-6, IL-8, and GM-CSF and inhibits hemopoiesis of myeloid progenitor cells in colony forming assays. Recombinant human IL-17D is a 40.5 kDa disulfide-linked homodimer of two 185 amino acid polypeptide chains.

Biochem/physiol Actions

IL-17D is a disulfide-linked homodimer of two 185 amino acid polypeptide chains. Recombinant human IL-17D is a 40.5 kDa disulfide-linked homodimer of two 185 amino acid polypeptide chains.

Sequence

APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP

Physical form

Lyophilized from 1x PBS, pH 7.2.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(2), 773-778 (2000-01-19)
IL-17 is a T cell-derived cytokine that may play an important role in the initiation or maintenance of the proinflammatory response. Whereas expression of IL-17 is restricted to activated T cells, the IL-17 receptor is found to be widely expressed
Juan Liao et al.
PloS one, 9(9), e108777-e108777 (2014-10-01)
The pathogenesis of complex diseases, such as type 1 diabetes (T1D), derives from interactions between host genetics and environmental factors. Previous studies have suggested that viral infection plays a significant role in initiation of T1D in genetically predisposed individuals. T1D
T A Moseley et al.
Cytokine & growth factor reviews, 14(2), 155-174 (2003-03-26)
Interleukin-17 (IL-17) is a pro-inflammatory cytokine secreted by activated T-cells. Recently discovered related molecules are forming a family of cytokines, the IL-17 family. The prototype member of the family has been designated IL-17A. Due to recent advances in the human
Shi Li et al.
Chinese medical journal, 128(14), 1932-1941 (2015-07-15)
Interleukin (IL)-27 has been reported to have anti-proliferate and anti-angiogenic activities on cancer cells. However, the involvement of IL-27 in malignant pleural effusion (MPE) remains unknown. Thus, in this research, we compared the immune functions of IL-27, interferon (IFN)-γ, and
Ming-Chieh Ma et al.
Basic research in cardiology, 110(3), 22-22 (2015-03-31)
Patients with coronary artery disease show high serum levels of interleukin (IL)-27, a novel member of the IL-6 family. However, the function of IL-27 in hearts suffering ischemia/reperfusion (IR) injury is unclear. Here, we showed increased expression of mRNA for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service