Merck
  • Improved performance of dye-sensitized solar cells using dual-function TiO(2) nanowire photoelectrode.
Improved performance of dye-sensitized solar cells using dual-function TiO(2) nanowire photoelectrode.
Optics express (2015-09-26)
Zico Alaia Akbar, Ji Hye Oh, Wisnu Tantyo Hadmojo, Su Ji Yang, Young Rag Do, Sung-Yeong Jang
ABSTRACT
A unique, hierarchically structured, aggregated TiO(2) nanowire (A-TiO(2)-nw) is prepared by solvothermal synthesis and used as a dual-functioning photoelectrode in dye-sensitized solar cells (DSSCs). The A-TiO(2)-nw shows improved light scattering compared to conventional TiO(2) nanoparticles (TiO(2)-np) and dramatically enhanced dye adsorption compared to conventional scattering particles (CSP). The A-TiO(2)-nw is used as a scattering layer for bilayer photoelectrodes (TiO(2)-np/A-TiO(2)-nw) in DSSCs to compare the cell performance to that of devices using state-of-the-art photoelectrode architectures (TiO(2)-np/CSP). The DSSCs fabricated using bilayers of TiO(2)-np/A-TiO(2)-nw show improved power conversion efficiency (9.1%) and current density (14.88 mA cm(-2)) compared to those using single-layer TiO(2)-np (7.6% and 11.84 mA cm(-2)) or TiO(2)-np/CSP bilayer structures (8.7% and 13.81 mA cm(-2)). The unique contribution of the A-TiO(2)-nw layers to the device performance is confirmed by studying the incident photon-to-current efficiency. The enhanced external quantum efficiencies at approximately 520 nm and 650 nm clearly reveal the dual functionality of A-TiO(2)-nw. These unique properties of A-TiO(2)-nw may be applied in other devices utilizing light-scattering n-type semiconductor.
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Lithium acetate dihydrate, BioUltra, ≥99.0% (anhydrous)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Lithium acetate dihydrate, BioXtra
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Sigma-Aldrich
Lithium acetate dihydrate, reagent grade