Merck
  • Amination of Aryl Halides and Esters Using Intensified Continuous Flow Processing.
Amination of Aryl Halides and Esters Using Intensified Continuous Flow Processing.
Molecules (Basel, Switzerland) (2015-10-02)
Thomas M Kohl, Christian H Hornung, John Tsanaktsidis
ABSTRACT
Significant process intensification of the amination reactions of aryl halides and esters has been demonstrated using continuous flow processing. Using this technology traditionally difficult amination reactions have been performed safely at elevated temperatures. These reactions were successfully conducted on laboratory scale coil reactor modules with 1 mm internal diameter (ID) and on a preparatory scale tubular reactor with 6 mm ID containing static mixers.
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Methyl 3-mercaptopropionate, 98%