Merck
  • MEK Inhibition Overcomes Cisplatin Resistance Conferred by SOS/MAPK Pathway Activation in Squamous Cell Carcinoma.
MEK Inhibition Overcomes Cisplatin Resistance Conferred by SOS/MAPK Pathway Activation in Squamous Cell Carcinoma.
Molecular cancer therapeutics (2015-05-06)
Li Ren Kong, Kian Ngiap Chua, Wen Jing Sim, Hsien Chun Ng, Chonglei Bi, Jingshan Ho, Min En Nga, Yin Huei Pang, Weijie Richard Ong, Ross Andrew Soo, Hung Huynh, Wee Joo Chng, Jean-Paul Thiery, Boon Cher Goh
ABSTRACT
Genomic analyses of squamous cell carcinoma (SCC) have yet to yield significant strategies against pathway activation to improve treatment. Platinum-based chemotherapy remains the mainstay of treatment for SCC of different histotypes either as a single-agent or alongside other chemotherapeutic drugs or radiotherapy; however, resistance inevitably emerges, which limits the duration of treatment response. To elucidate mechanisms that mediate resistance to cisplatin, we compared drug-induced perturbations to gene and protein expression between cisplatin-sensitive and -resistant SCC cells, and identified MAPK-ERK pathway upregulation and activation in drug-resistant cells. ERK-induced resistance appeared to be activated by Son of Sevenless (SOS) upstream, and mediated through Bim degradation downstream. Clinically, elevated p-ERK expression was associated with shorter disease-free survival in patients with locally advanced head and neck SCC treated with concurrent chemoradiation. Inhibition of MEK/ERK, but not that of EGFR or RAF, augmented cisplatin sensitivity in vitro and demonstrated efficacy and tolerability in vivo. Collectively, these findings suggest that inhibition of the activated SOS-MAPK-ERK pathway may augment patient responses to cisplatin treatment.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sos2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bcl2l11
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Map2k1
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK3
Sigma-Aldrich
MISSION® esiRNA, targeting human SOS1
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP2K1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Src
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sos1
Sigma-Aldrich
MISSION® esiRNA, targeting human SOS2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mapk3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Map2k2
Sigma-Aldrich
BICINE, BioXtra, ≥99% (titration)
Sigma-Aldrich
BICINE, ≥99% (titration)