Skip to Content
Merck
All Photos(4)

Key Documents

B3876

Sigma-Aldrich

BICINE

≥99% (titration)

Synonym(s):

N,N-Bis(2-hydroxyethyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4
CAS Number:
Molecular Weight:
163.17
Beilstein:
1769362
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (titration)

form

powder or crystals

pH

3.5-5.0

useful pH range

7.6-9.0

pKa (25 °C)

8.3

solubility

water: 25 % (w/w), clear, colorless

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

OCCN(CCO)CC(O)=O

InChI

1S/C6H13NO4/c8-3-1-7(2-4-9)5-6(10)11/h8-9H,1-5H2,(H,10,11)

InChI key

FSVCELGFZIQNCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Recommended buffer for low temperature biochemical work. Used in the preparation of stable substrate solution for serum guanase determination. Spacer in plasma protein fractionation with Sephadex® by isotachophoresis.

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The myosin dimer: an intermediate in the self-assembly of the thick filament of vertebrate skeletal muscle.
J S Davis et al.
FEBS letters, 140(2), 293-297 (1982-04-19)
T A Churchill et al.
Transplantation, 65(4), 551-559 (1998-03-21)
This study was designed to investigate the effects of a modified University of Wisconsin (UW) solution supplemented with one of four buffering agents (histidine, bicine [N,N-bis(2-hydroxyethyl)glycine], tricine [N-tris(hydroxymethyl)methylglycine], and Tris) on liver metabolism during cold ischemic storage. Rat livers were
R Nakon et al.
Science (New York, N.Y.), 221(4612), 749-750 (1983-08-19)
Metal-ion affinity (formation) constants were determined for two "Good's" buffers, N-tris(hydroxymethyl)methyl-2-aminoethanesulfonic acid (TES) and N,N-bis(2-hydroxyethyl)glycine (bicine). The metal chelates formed undergo loss of an internal ligand (alcohol) proton (bicine) and undergo hydrolysis (bicine and TES) and dimerization reactions (TES). Bicine
Irradiation inactivation analysis of acetylcholinesterase and the effect of buffer salts.
D Parkinson et al.
Radiation research, 90(2), 252-259 (1982-05-01)
Raewyn M Town et al.
The journal of physical chemistry. A, 112(12), 2563-2571 (2008-03-04)
The impact of ligand protonation on the complexation kinetics of higher-order complexes is quantitatively described. The theory is formulated on the basis of the usual situation for metal complex formation in aqueous systems in which the exchange of water for

Protocols

Objective: To standardize a procedure for the recycling micro-assay of β-NAD and β-NADH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service