Merck
  • High Sensitivity RT-qPCR Assay of Nonlabeled siRNA in Small Blood Volume for Pharmacokinetic Studies: Application to Survivin siRNA.
High Sensitivity RT-qPCR Assay of Nonlabeled siRNA in Small Blood Volume for Pharmacokinetic Studies: Application to Survivin siRNA.
The AAPS journal (2015-08-20)
Bertrand Z Yeung, Ze Lu, Guillaume M Wientjes, Jessie L-S Au
ABSTRACT
RNAi therapeutics provide an opportunity to correct faulty genes, and several RNAi have entered clinical evaluation. The existing quantification methods typically use radioactivity- or fluorescence-labeled RNAi, require large blood volumes, and/or have a limited dynamic detection range. We established a quantitative reverse transcriptase real-time polymerase chain reaction (RT-qPCR) assay to measure RNAi; the model analyte was survivin siRNA (siSurvivin). A second siRNA was used as the internal standard. The three major steps were (a) extraction of the two siRNAs from blood or water, (b) synthesis of their cDNA by poly-A extension, and (c) qPCR of cDNA. Standard curves were established. Utility of the assay was demonstrated in a pharmacokinetic study where all 12 samples for the blood concentration-time profile were obtained from a single mouse given an intravenous dose of 1 nmole siSurvivin (prepared as lipoplex with pegylated cationic liposomes). The RT-qPCR assay was sensitive (lower detection limit of 100 fM) and had a 5 × 107-fold dynamic range and low sample volume requirement (10 μL). The 16-point standard curves constructed using whole blood samples were linear (R (2) > 0.98). The intraday and interday variations for the slopes were ≤6%, although the variations for accuracy and precision at individual concentrations were substantially higher (58-145%). Standard curves prepared with water in place of blood showed similar results (<6% difference), indicating water may be used when blood is not available. The current RT-qPCR assay enabled the measurement of nonlabeled siRNA in small volume of blood samples.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥99.0% (10 mg phospholipid per ml CHCl3, TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥97.0% (TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Birc5
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis