Merck
  • Fundamentals of MOF Thin Film Growth via Liquid-Phase Epitaxy: Investigating the Initiation of Deposition and the Influence of Temperature.
Fundamentals of MOF Thin Film Growth via Liquid-Phase Epitaxy: Investigating the Initiation of Deposition and the Influence of Temperature.
Langmuir : the ACS journal of surfaces and colloids (2015-05-29)
Monica L Ohnsorg, Christopher K Beaudoin, Mary E Anderson
ABSTRACT
Thin films can integrate the versatility and great potential found in the emerging field of metal-organic frameworks directly into device architectures. For fabrication of smart interfaces containing surface-anchored metal-organic frameworks, it is important to understand how the foundational layers form to create the interface between the underlying substrate and porous framework. Herein, the formation and morphology of the first ten cycles of film deposition are investigated for the well-studied HKUST-1 system. Effects of processing variables, such as deposition temperature and substrate quality, are studied. Sequences of scanning probe microscopy images collected after cycles of alternating solution-phase deposition reveal the formation of a discontinuous surface with nucleating and growing crystallites consistent with a Volmer-Weber growth mechanism. Quantitative image analysis determines surface roughness and surface coverage as a function of deposition cycles, producing insight regarding growth and structure of foundational film layers. For carboxylic acid terminated self-assembled monolayers on gold, preferred crystal orientation is influenced by deposition temperature with crystal growth along [100] observed at 25 °C and [111] favored at 50 °C. This difference in crystal orientation results in reduced surface roughness and increased surface coverage at 50 °C. To properly fabricate and fully determine the potential of this material for industrial applications, fundamental understanding of film formation is crucial.
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
16-Mercaptohexadecanoic acid, 98%
Sigma-Aldrich
Copper(II) acetate hydrate, 98%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
16-Mercaptohexadecanoic acid, 90%
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
Copper(II) acetate monohydrate, 99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes