Merck
  • Notch1 activity in the olfactory bulb is odour-dependent and contributes to olfactory behaviour.
Notch1 activity in the olfactory bulb is odour-dependent and contributes to olfactory behaviour.
The European journal of neuroscience (2014-09-23)
Emanuele Brai, Swananda Marathe, Lorena Zentilin, Mauro Giacca, Johannes Nimpf, Robert Kretz, Alessandra Scotti, Lavinia Alberi
ABSTRACT
Notch signalling plays an important role in synaptic plasticity, learning and memory functions in both Drosophila and rodents. In this paper, we report that this feature is not restricted to hippocampal networks but also involves the olfactory bulb (OB). Odour discrimination and olfactory learning in rodents are essential for survival. Notch1 expression is enriched in mitral cells of the mouse OB. These principal neurons are responsive to specific input odorants and relay the signal to the olfactory cortex. Olfactory stimulation activates a subset of mitral cells, which show an increase in Notch activity. In Notch1cKOKln mice, the loss of Notch1 in mitral cells affects the magnitude of the neuronal response to olfactory stimuli. In addition, Notch1cKOKln mice display reduced olfactory aversion to propionic acid as compared to wildtype controls. This indicates, for the first time, that Notch1 is involved in olfactory processing and may contribute to olfactory behaviour.
Product Description
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Pentyl acetate, puriss. p.a., ≥98.5% (GC)
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Amyl acetate, ≥99%, FG
Sigma-Aldrich
2-Methylbutyric acid, 98%
Sigma-Aldrich
2-Methylbutyric acid, ≥98%, FG
Sigma-Aldrich
Pentyl acetate, 99%
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse
Supelco
Pentyl acetate, analytical standard
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
(±)-2-Methylbutyric acid, natural, ≥98%, FG
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Propionic acid, ≥99.5%