Skip to Content
Merck
All Photos(3)

Key Documents

W212717

Sigma-Aldrich

Benzaldehyde

greener alternative

natural, FCC, FG

Synonym(s):

Bitter almond

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHO
CAS Number:
Molecular Weight:
106.12
FEMA Number:
2127
Beilstein:
471223
EC Number:
Council of Europe no.:
101
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39023701
PubChem Substance ID:
Flavis number:
5.013
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

3.7 (vs air)

vapor pressure

4 mmHg ( 45 °C)

Assay

≥98%

form

liquid

autoignition temp.

374 °F

composition

contains IFRA restricted benzaldehyde

expl. lim.

1.4 %, 20 °F

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.545 (lit.)

pH

5.9 (20 °C)

bp

178-179 °C (lit.)

mp

−26 °C (lit.)

density

1.044 g/cm3 at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

benzaldehyde

greener alternative category

Organoleptic

almond; cherry; sweet

SMILES string

O=Cc1ccccc1

InChI

1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H

InChI key

HUMNYLRZRPPJDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzaldehyde is an aromatic flavor compound with a characteristic bitter almond taste and odor.[1] It has been identified in the volatile fractions of mushroom[2], medlar seeds[3] and cheese.[4]
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenaroli G
Fenaroli's Handbook of Flavor Ingredients, 40-40 (1971)
Conversion of phenylalanine to benzaldehyde initiated by an aminotransferase in Lactobacillus plantarum
Groot MNN & de Bont JA.
Applied and Environmental Microbiology, 64(8), 3009-3013 (1998)
Supercritical fluid extraction of volatile components from Bunium persicum Boiss.(black cumin) and Mespilus germanica L.(medlar) seeds
Pourmortazavi SM, et al.
J. Food Compos. Anal., 18(5), 439-446 (2005)
Volatile components of mushroom (Agaricus subrufecens)
Chen CC & Wu CM
Journal of Food Science, 49(4), 1208-1209 (1984)
Longyun Lv et al.
ACS combinatorial science, 15(4), 183-192 (2013-02-22)
We previously reported the novel efficient proton/heat-promoted four-component reactions (4CRs) of but-2-ynedioates, two same/different primary amines, and aldehydes for the synthesis of tetra- and pentasubstituted polyfunctional dihydropyrroles. If aromatic and aliphatic amines were used as reagents, four different series of

Questions

  1. We need to know the packsize of sample-K, please tell us from W212717-SAMPLE-K

    1 answer
    1. The SAMPLE-K package size for this item is 10g.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service