Merck
  • Folding graft copolymer with pendant drug segments for co-delivery of anticancer drugs.
Folding graft copolymer with pendant drug segments for co-delivery of anticancer drugs.
Biomaterials (2014-05-31)
Wanyi Tai, Ran Mo, Yue Lu, Tianyue Jiang, Zhen Gu
ABSTRACT
A graft copolymer with pendant drug segments can fold into nanostructures in a protein folding-like manner. The graft copolymer is constructed by directly polymerizing γ-camptothecin-glutamate N-carboxyanhydride (Glu(CPT)-NCA) on multiple sites of poly(ethylene glycol) (PEG)-based main chain via the ring open polymerization (ROP). The "purely" conjugated anticancer agent camptothecin (CPT) is hydrophobic and serves as the principal driving force during the folding process. When exposed to water, the obtained copolymer, together with doxorubicin (Dox), another anticancer agent, can fold into monodispersed nanocarriers (with a diameter of around 50 nm) for dual-drug delivery. Equipped with a PEG shell, the nanocarriers displayed good stability and can be internalized by a variety of cancer cell lines via the lipid raft and clathrin-mediated endocytotic pathway without premature leakage, which showed a high synergetic activity of CPT and Dox toward various cancer cells. In vivo study validated that the nanocarriers exhibited strong accumulation in tumor sites and showed a prominent anticancer activity against the lung cancer xenograft mice model compared with free drugs.
Sigma-Aldrich
Acetamide, sublimed, 99%
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Di-tert-butyl dicarbonate, ≥98.0% (GC)
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, ≥99%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Tris(2-aminoethyl)amine, 96%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, 99%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Boc-Glu-OtBu
Sigma-Aldrich
Acetamide, ~99% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Acetamide, ≥99.0% (GC)
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in THF
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in methylene chloride
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Triphosgene, reagent grade, 98%
Sigma-Aldrich
DIC, purum, ≥98.0% (GC)
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 1.0 M in THF
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
DIC, 99%