Skip to Content
Merck
All Photos(1)

Key Documents

15217

Sigma-Aldrich

Bis(trichloromethyl) carbonate

purum, ≥99.0% (AT)

Synonym(s):

Triphosgene, tri-Phosgene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3COCOOCCl3
CAS Number:
Molecular Weight:
296.75
Beilstein:
1787583
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
≥99.0% (AT)

grade

purum

Quality Level

Assay

≥99.0% (AT)

form

solid

mp

77-82 °C

functional group

carbonate
chloro

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)OC(=O)OC(Cl)(Cl)Cl

InChI

1S/C3Cl6O3/c4-2(5,6)11-1(10)12-3(7,8)9

InChI key

UCPYLLCMEDAXFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(trichloromethyl) carbonate was used in the preparation of Fmoc-amino acid chlorides. It was also used in the preparation of aryl-(Z)-vinyl chlorides.

Other Notes

Crystalline, easy-to-handle substitute for phosgene; 1 mole of the reagent corresponds in its reactivity to 3 moles phosgene

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Cortez et al.
Synthetic Communications, 21, 285-285 (1991)
G.B. Gill et al.
Tetrahedron Letters, 30, 3229-3229 (1989)
H. Eckert, B. Forster
Angewandte Chemie (International Edition in English), 99, 922-922 (1987)
P.R.M. Muller
Spec. Chem., 14, 357-357 (1994)
R M Cicchillo et al.
Carbohydrate research, 328(3), 431-434 (2000-11-10)
Treating partially protected sugar hemiacetals with triphosgene in THF results in the formation of glycosyl chlorides. The method is compatible with acid-sensitive isopropylidene protecting groups in the hemiacetal substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service